Skip to Content
Merck
All Photos(1)

Key Documents

C0990000

Cetyl alcohol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Hexadecanol, Cetyl alcohol, Palmityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
Beilstein:
1748475
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

8.34 (vs air)

vapor pressure

<0.01 mmHg ( 43 °C)

API family

cetyl alcohol

autoignition temp.

483 °F

expl. lim.

8 %

manufacturer/tradename

EDQM

bp

179-181 °C/10 mmHg (lit.)

mp

48-50 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCCCCCO

InChI

1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

BXWNKGSJHAJOGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cetyl alcohol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Dashti et al.
Chemosphere, 70(3), 475-479 (2007-08-07)
Bacteria and fungi in pristine and oily desert soil samples were counted on inorganic medium aliquots containing 0.5% hexadecane, hexadecanol, hexadecanal or hexadecanoic acid, as sole sources of carbon and energy. It was found that the carbon and energy source
Kerstin Wagner et al.
Chemistry and physics of lipids, 145(2), 119-127 (2006-12-26)
The monolayer structure of pure dipalmitoylphosphatidylcholine (DPPC) and equimolar mixtures of DPPC/n-hexadecanol (C(16)OH) and DPPC/dipalmitoylglycerol (DPG) are studied by the film balance technique and grazing incidence X-ray diffraction measurements. At 20 degrees C, the binary systems exhibit complete miscibility. In
Flávia C Carvalho et al.
Journal of pharmaceutical sciences, 99(5), 2367-2374 (2009-12-08)
The development of a controlled-release dosage form of zidovudine (AZT) is of crucial importance, in view of the pharmacokinetics of its toxic activity. A suitable drug delivery system could increase AZT bioavailability, reducing its dose-dependent side effects. In this study
Laurent Perge et al.
International journal of pharmaceutics, 422(1-2), 59-67 (2011-10-27)
A hot melt dispersion method was used to prepare new sustained release ibuprofen composite microparticles of a solid lipid at ambient temperature, cetyl alcohol. The dispersion of colloidal silicon dioxide nanoparticles (hydrophilic Aerosil 200 or hydrophobic Aerosil R974) either in
Dhanalakshmi S Anbukumar et al.
Journal of lipid research, 51(5), 1085-1092 (2009-12-19)
Neutrophils are important in the host response against invading pathogens. One chemical defense mechanism employed by neutrophils involves the production of myeloperoxidase (MPO)-derived HOCl. 2-Chlorohexadecanal (2-ClHDA) is a naturally occurring lipid product of HOCl targeting the vinyl ether bond of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service