Skip to Content
Merck
All Photos(1)

Key Documents

C0685000

Cefotaxime sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cefotaxime sodium salt, Cefotaxim sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N5NaO7S2
CAS Number:
Molecular Weight:
477.45
Beilstein:
5711411
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefotaxime

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[H][C@]12SCC(COC(C)=O)=C(N1C(=O)[C@H]2NC(=O)C(=N/OC)\c3csc(N)n3)C([O-])=O

InChI

1S/C16H17N5O7S2.Na/c1-6(22)28-3-7-4-29-14-10(13(24)21(14)11(7)15(25)26)19-12(23)9(20-27-2)8-5-30-16(17)18-8;/h5,10,14H,3-4H2,1-2H3,(H2,17,18)(H,19,23)(H,25,26);/q;+1/p-1/b20-9-;/t10-,14-;/m1./s1

InChI key

AZZMGZXNTDTSME-JUZDKLSSSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefotaxime sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cefotaxim inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs) which inhibits the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls. As a result, bacteria lyse due to cell wall autolytic enzymes.
Mode of Action: Inhibits bacterial cell wall synthesis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Broad spectrum third generation cephalosporin antibiotic.
Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Asif et al.
Plant cell reports, 32(10), 1637-1646 (2013-07-31)
Cefotaxime (100 mg/l) mitigate occasional gram negative bacterial contamination in wheat and triticale microspore culture and most importantly it increases cell growth and green plant production. Isolated microspore culture is a promising option to rapidly fix the product of meiotic recombination
Elena Ojer-Usoz et al.
Meat science, 93(2), 316-321 (2012-10-16)
Patterns of resistance in β-lactamase-producing Enterobacteriaceae family were investigated in isolates from 141 meat products (beef, poultry and pork) purchased in Spain. The strains that grow in ChromID ESBL agar plates were confirmed using the paired disk diffusion method. Resistance
Ryuichi Nakano et al.
Journal of medical microbiology, 61(Pt 12), 1727-1735 (2012-09-01)
Proteus mirabilis is a common cause of urinary tract infection. Wild-type P. mirabilis strains are usually susceptible to penicillins and cephalosporins, but occurrences of P. mirabilis producing extended-spectrum β-lactamases (ESBLs) have been recently reported. Here, we surveyed the prevalence of
Martijn F Schenk et al.
PLoS genetics, 8(6), e1002783-e1002783 (2012-07-05)
For a quantitative understanding of the process of adaptation, we need to understand its "raw material," that is, the frequency and fitness effects of beneficial mutations. At present, most empirical evidence suggests an exponential distribution of fitness effects of beneficial
I Mounchetrou Njoya et al.
Euro surveillance : bulletin Europeen sur les maladies transmissibles = European communicable disease bulletin, 17(34) (2012-09-04)
In April 2012, a cluster of two cases of meningococcal disease caused by rifampicin-resistant C meningococci was reported in the Champagne-Ardenne region, France. The two cases occurred in a student population living in the same town but studying at different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service