Skip to Content
Merck
All Photos(1)

Documents

C0430

Supelco

(−)-Cotinine solution

drug standard, 1.0 mg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

drug standard

Quality Level

Agency

EPA 1694

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

CN1[C@@H](CCC1=O)c2cccnc2

InChI

1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/t9-/m0/s1

InChI key

UIKROCXWUNQSPJ-VIFPVBQESA-N

Gene Information

General description

(−)-Cotinine is a major metabolite of nicotine, widely used as a biochemical marker for to evaluate the exposure to environmental tobacco smoke, for the validation of abstinence smoking and to determine the percentage of active smoking behavior.

Application

(−)-Cotinine solution has been used as a standard in the analysis of nicotine in air samples using ultra performance liquid chromatography-mass spectrometry (UPLC-MS). (−)-Cotinine may be may used as an analytical reference standard for the quantification of the analyte in biological samples using high-performance liquid chromatography technique.
(−)-Cotinine solution has been used as a standard in the analysis of nicotine in air samples using ultra performance liquid chromatography-mass spectrometry (UPLC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mohammad A Kaisar et al.
BMC neuroscience, 18(1), 71-71 (2017-10-13)
A sensitive, rapid and selective UHPLC-MS/MS method has been developed and validated for the quantification of Nicotine (NT) and Cotinine (CN) using Continine-d A linear response function was established for the range of concentrations 3-200 (r > 0.995) for NT and 3-600 ng/mL
A simple, sensitive, and rapid method for the determination of cotinine in urine by high-performance liquid chromatography with UV detection
Rabbaa-Khabbaz L, et al.
Journal of Chromatographic Science, 44(9), 535-538 (2006)
Rapid and sensitive high-performance liquid chromatographic determination of nicotine and cotinine in nonsmoker human and rat urines
Oddoze.C, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 708(1-2), 95-101 (1998)
Assessment of nicotine for second hand smoke exposure in some public places in Romania by UPLC-MS
Bratan A, et al.
Chemistry Central Journal, 8(1), 29-29 (2014)
A sensitive HPLC-ESI-MS-MS method for the determination of cotinine in urine
Apinan R, et al.
Journal of Chromatographic Science, 48(6), 460-465 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service