Skip to Content
Merck
All Photos(1)

Key Documents

C0200000

Calcitonin salmon

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Thyrocalcitonin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C145H240N44O48S2
CAS Number:
Molecular Weight:
3431.85
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

calcitonin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H]N[C@H]1CSSC[C@H](NC(=O)[C@@H](NC(=O)[C@H](CO)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CO)NC1=O)C(C)O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc2cnc[nH]2)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](Cc3ccc(O)cc3)C(=O)N4CCC[C@H]4C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C(C)O)C(=O)NCC(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H](C(C)O)C(=O)N5CCC[C@H]5C(N)=O

InChI

1S/C145H240N44O48S2/c1-65(2)45-86(175-139(232)110(70(11)12)183-136(229)99-63-239-238-62-79(148)117(210)178-96(59-191)134(227)174-92(52-104(151)202)131(224)172-90(49-69(9)10)129(222)180-98(61-193)135(228)187-114(74(16)197)142(235)181-99)118(211)158-55-106(204)162-80(25-18-20-40-146)120(213)169-89(48-68(7)8)128(221)179-97(60-192)133(226)167-83(34-37-102(149)200)122(215)165-85(36-39-109(207)208)123(216)171-88(47-67(5)6)127(220)173-91(51-77-54-156-64-161-77)130(223)164-81(26-19-21-41-147)121(214)170-87(46-66(3)4)126(219)166-84(35-38-103(150)201)125(218)186-113(73(15)196)141(234)177-94(50-76-30-32-78(199)33-31-76)143(236)189-44-24-29-101(189)137(230)168-82(27-22-42-157-145(154)155)124(217)185-112(72(14)195)140(233)176-93(53-105(152)203)132(225)184-111(71(13)194)138(231)160-56-107(205)163-95(58-190)119(212)159-57-108(206)182-115(75(17)198)144(237)188-43-23-28-100(188)116(153)209/h30-33,54,64-75,79-101,110-115,190-199H,18-29,34-53,55-63,146-148H2,1-17H3,(H2,149,200)(H2,150,201)(H2,151,202)(H2,152,203)(H2,153,209)(H,156,161)(H,158,211)(H,159,212)(H,160,231)(H,162,204)(H,163,205)(H,164,223)(H,165,215)(H,166,219)(H,167,226)(H,168,230)(H,169,213)(H,170,214)(H,171,216)(H,172,224)(H,173,220)(H,174,227)(H,175,232)(H,176,233)(H,177,234)(H,178,210)(H,179,221)(H,180,222)(H,181,235)(H,182,206)(H,183,229)(H,184,225)(H,185,217)(H,186,218)(H,187,228)(H,207,208)(H4,154,155,157)/t71-,72-,73-,74-,75-,79+,80+,81+,82+,83+,84+,85+,86+,87+,88+,89+,90+,91+,92+,93+,94+,95+,96+,97+,98+,99+,100+,101+,110+,111+,112+,113+,114+,115+/m1/s1

InChI key

BBBFJLBPOGFECG-VJVYQDLKSA-N

Gene Information

human ... CALCR(799)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2 [Disulfide bridge: 1-7]

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Calcitonin salmon EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Hypocalcemic hormone produced by the parafollicular C cells of the thyroid or by the ultimobranchial bodies of nonmammalian vertebrates. Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

32 amino acid polypeptide, 8 of which are conserved across all species.
Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathew W Jones et al.
Journal of the American Chemical Society, 134(17), 7406-7413 (2012-04-13)
Direct polymer conjugation at peptide tyrosine residues is described. In this study Tyr residues of both leucine enkephalin and salmon calcitonin (sCT) were targeted using appropriate diazonium salt-terminated linear monomethoxy poly(ethylene glycol)s (mPEGs) and poly(mPEG) methacrylate prepared by atom transfer
Mingshi Yang et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 47(1), 235-243 (2012-06-12)
The aim of this study was to investigate two types of chitosan-modified poly (DL-lactic-co-glycolic acid) (PLGA) nanocomposite particles containing salmon calcitonin for pulmonary delivery, which were obtained using spray drying fluidized bed granulation (Agglomaster™) and dry powder coating techniques (Mechanofusion™)
Neil Binkley et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 27(8), 1821-1829 (2012-03-23)
The Oral Calcitonin in Postmenopausal Osteoporosis (ORACAL) study was a randomized, double-blind, double-dummy, active- and placebo-controlled, multiple-dose, phase 3 study to assess the efficacy and safety of oral recombinant calcitonin for treatment of postmenopausal osteoporosis. A total of 565 women
Leonie Baginski et al.
Pharmaceutical research, 29(6), 1425-1434 (2012-02-11)
To investigate DSPE-PEG(2000)-based micellar formulations of salmon calcitonin (sCT) for their ability to improve pulmonary delivery. Micelles were characterised by DLS and (31)P-NMR spectroscopy. Stability against sCT degrading peptidases, trypsin, α-chymotrypsin and neutrophil elastase as well as their influence on
Viswatej Vemulapalli et al.
Journal of pharmaceutical sciences, 101(8), 2861-2869 (2012-05-31)
The purpose of this study was to determine the effect of microneedle (MN) technology and its combination with iontophoresis (ITP) on the in vivo transdermal delivery of salmon calcitonin (sCT). Maltose MNs (500 µm) were used to porate skin prior

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service