Skip to Content
Merck
All Photos(1)

Key Documents

BCR089

Ti Al6 V4 (Al, V)

BCR®, certified reference material, disc

Synonym(s):

Titanium-aluminum-vanadium alloy, Aluminum-titanium-vanadium alloy Al6TiV

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

form

disc

manufacturer/tradename

JRC

format

matrix material

SMILES string

[Al].[Ti].[V]

InChI

1S/Al.Ti.V

InChI key

GDAKJZDZIXHMSC-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR089

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service