Skip to Content
Merck
All Photos(1)

Key Documents

B1045000

Betamethasone sodium phosphate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Betamethasone 21-phosphate disodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28FNa2O8P
CAS Number:
Molecular Weight:
516.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

betamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[Na+].[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@H](C)[C@]4(O)C(=O)COP([O-])([O-])=O

InChI

1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15-,16-,17-,19-,20-,21-,22-;;/m0../s1

InChI key

PLCQGRYPOISRTQ-LWCNAHDDSA-L

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betamethasone sodium phosphate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Diagnostic Implications

Betamethasone is a corticosteroid with anti-inflammatory and immunosuppressive abilities, used especially where water retention is undesirable. It is applied as a topical cream, ointment, lotion or gel to treat itching (e.g. from eczema). Corticosteroid used to stimulate fetal lung maturation and to decrease the incidence and mortality from intracranial hemorrhage in premature infants

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B - STOT RE 2

Target Organs

Liver,Kidney,Endocrine system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jun Lu et al.
Journal of chromatography. A, 1217(44), 6932-6941 (2010-09-22)
Betamethasone Sodium Phosphate and Betamethasone Acetate are the two corticosteroids active pharmaceutical ingredients (APIs) that are present in the injectable formulation, Celestone Chronodose(®) Injection. It is extremely challenging to develop a Quality Control friendly HPLC method to separate all the
Ibrahim Coban et al.
Foot and ankle surgery : official journal of the European Society of Foot and Ankle Surgeons, 15(1), 22-27 (2009-02-17)
To determine the effects of human amniotic fluid and membrane in the treatment of Achilles tendon ruptures, 72 tendons of 36 Wistar rats were injected with betamethasone sodium phosphate. By the end of fourth week, both tendons were tenotomized and
Masatomo Kato et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 27(4), 353-360 (2011-05-18)
Glucocorticoids exert their actions via the glucocorticoid receptor through at least 2 intracellular mechanisms, known as transrepression and transactivation. It has been hypothesized that transrepression is the basis of their anti-inflammatory effects, whereas transactivation has been assumed to cause their
Tsutomu Sakai et al.
Investigative ophthalmology & visual science, 52(3), 1516-1521 (2010-12-24)
The therapeutic effects of betamethasone phosphate (BP) encapsulated in biocompatible and biodegradable blended nanoparticles of poly(lactic acid) (PLA) homopolymers and PEG-block-PLA copolymers (stealth nanosteroids) were examined in an experimental autoimmune uveoretinitis (EAU) model in Lewis rats. EAU was induced by
Istemi Yucel et al.
Archives of orthopaedic and trauma surgery, 129(5), 695-701 (2008-10-08)
The aim of the study was to compare the efficacies of steroid injections guided by scintigraphy, ultrasonography, and palpation in plantar fasciitis. A total of 35 heels of 27 patients were randomly assigned to three steroid injection groups: palpation-guided (pg)

Articles

Development of an HPTLC based method, using Silica Gel 60 F254 plate, to determine Betamethasone Sodium Phosphate over a concentration range of 1 to 7 µg/spot.

Development of an HPTLC based method, using Silica Gel 60 F254 plate, to determine Betamethasone Sodium Phosphate over a concentration range of 1 to 7 µg/spot.

Development of an HPTLC based method, using Silica Gel 60 F254 plate, to determine Betamethasone Sodium Phosphate over a concentration range of 1 to 7 µg/spot.

Development of an HPTLC based method, using Silica Gel 60 F254 plate, to determine Betamethasone Sodium Phosphate over a concentration range of 1 to 7 µg/spot.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service