Skip to Content
Merck
All Photos(1)

Key Documents

A0365900

Amikacin sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Amikacin disulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43N5O13 · 2H2SO4
CAS Number:
Molecular Weight:
781.76
Beilstein:
6172633
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

amikacin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O

InChI

1S/C22H43N5O13.2H2O4S/c23-2-1-8(29)20(36)27-7-3-6(25)18(39-22-16(34)15(33)13(31)9(4-24)37-22)17(35)19(7)40-21-14(32)11(26)12(30)10(5-28)38-21;2*1-5(2,3)4/h6-19,21-22,28-35H,1-5,23-26H2,(H,27,36);2*(H2,1,2,3,4)/t6-,7+,8-,9+,10+,11-,12+,13+,14+,15-,16+,17-,18+,19-,21+,22+;;/m0../s1

InChI key

FXKSEJFHKVNEFI-GCZBSULCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amikacin sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamal M Matar et al.
BioMed research international, 2013, 202818-202818 (2013-03-14)
Amikacin pharmacokinetic data in Kuwaiti (Arab) intensive care unit (ICU) patients are lacking. Fairly sparse serum amikacin peak and trough concentrations data were obtained from adult Kuwaiti ICU patients. The data were analysed using a nonparametric adaptive grid (NPAG) maximum
Kenneth S Chen et al.
Pediatric blood & cancer, 60(11), 1772-1777 (2013-06-22)
Children undergoing cancer therapy often receive aminoglycosides to treat febrile neutropenia or gram-negative infections. The magnitude of the risk of developing aminoglycoside-induced ototoxicity and the dose threshold at which that risk significantly increases are unknown. Eligible cancer patients received the
Adi V Gundlapalli et al.
Diagnostic microbiology and infectious disease, 75(3), 256-259 (2013-01-12)
Antimicrobial shortages have made treating certain infections more difficult. A web-based survey asking about experience with antimicrobial drug shortages was distributed in 2011 to 1328 infectious diseases physician members of the Emerging Infectious Diseases Network of the Infectious Diseases Society
Sabine Ladrech et al.
The European journal of neuroscience, 38(6), 2962-2972 (2013-07-10)
To examine whether an inflammatory process occurs in the amikacin-poisoned cochlea, we investigated the presence of the cytokines tumour necrosis factor-α (TNF-α), interleukin (IL)-1β, and IL-10. No TNF-α, IL-1β or IL-10 was detected in the cochlear perilymph after the loss
Anna Belcarz et al.
International journal of pharmaceutics, 454(1), 285-295 (2013-07-16)
Following the quest for new composite materials for bone tissue engineering, a novel elastic hydroxyapatite-glucan composite loaded with two aminoglycoside antibiotics was prepared. The porosity of the composite and the drug release profiles in closed-loop and semi-open systems were tested.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service