Skip to Content
Merck
All Photos(2)

Key Documents

97013

Supelco

trans-Cinnamic acid

analytical standard

Synonym(s):

(2E)-3-Phenyl-2-propenoic acid, trans-3-Phenylacrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O2
CAS Number:
Molecular Weight:
148.16
Beilstein:
1905952
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

133 °C (lit.)
133-137 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\c1ccccc1

InChI

1S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H,10,11)/b7-6+

InChI key

WBYWAXJHAXSJNI-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-Cinnamic acid occurs in plants, formed via deamination of L-phenylalanine in the presence of enzymatic catalyst, L-phenylalanine ammonia-lyase.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum curcuma

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of L-phenylalanine from trans-cinnamic acid with Rhodotorula glutinis containing L-phenylalanine ammonia-lyase activity.
Yamada S, et al.
Applied and Environmental Microbiology, 42(5), 773-778 (1981)
Salicylic Acid: A Multifaceted Hormone (2017)
Hee Jin Seo et al.
Journal of nanoscience and nanotechnology, 13(3), 1727-1732 (2013-06-13)
Novel liposomes composed of dioleoylphosphatidylethanolamine (DOPE) and 2-(hexadecyloxy) cinnamic acid (HOCA) were prepared by a detergent removal method. When the molar ratio of DOPE to HOCA was 4:1, 3:2, and 2:3, the florescence quenching degree of 5(6)-carboxylic fluorescein (CF, a
Bianca C Pérez et al.
Journal of medicinal chemistry, 56(2), 556-567 (2013-01-01)
The control of malaria is challenged by drug resistance, and new antimalarial drugs are needed. New drug discovery efforts include consideration of hybrid compounds as potential multitarget antimalarials. Previous work from our group has demonstrated that hybrid structures resulting from
Stanislava Gorjanović et al.
Journal of agricultural and food chemistry, 60(38), 9573-9580 (2012-09-07)
Hydrogen peroxide scavenging (HPS) activity of unfermented (green, yellow, and white), partially fermented (oolong), and completely fermented (black) tea ( Camellia sinensis ), maté ( Ilex paraguariensis ), and various herbal infusions, as well as individual compounds (flavan-3-ols, flavonols, cinnamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service