Skip to Content
Merck
All Photos(2)

Key Documents

94815

Supelco

Glycolic acid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Hydroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2COOH
CAS Number:
Molecular Weight:
76.05
Beilstein:
1209322
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas (Argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

75-80 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

OCC(O)=O

InChI

1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)

InChI key

AEMRFAOFKBGASW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>572.0 °F - (decomposition)

Flash Point(C)

> 300 °C - (decomposition)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thea R Pick et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(8), 3185-3190 (2013-02-06)
Photorespiratory carbon flux reaches up to a third of photosynthetic flux, thus contributes massively to the global carbon cycle. The pathway recycles glycolate-2-phosphate, the most abundant byproduct of RubisCO reactions. This oxygenation reaction of RubisCO and subsequent photorespiration significantly limit
W H Porter et al.
Journal of toxicology. Clinical toxicology, 39(6), 607-615 (2002-01-05)
To correlate serum glycolic acid levels with clinical severity and outcome in ethylene glycol poisoning and to determine if glycolic acid levels are predictive of renal failure and the need for hemodialysis. We measured serum ethylene glycol and glycolic acid
Ingela Bjurhager et al.
Biomacromolecules, 13(8), 2521-2527 (2012-07-10)
In 1628, the Swedish warship Vasa capsized on her maiden voyage and sank in the Stockholm harbor. The ship was recovered in 1961 and, after polyethylene glycol (PEG) impregnation, it was displayed in the Vasa museum. Chemical investigations of the
Genqiang Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 24(34), 4609-4613 (2012-06-26)
Novel ZnMn(2)O(4) ball-in-ball hollow microspheres are fabricated by a facile two-step method involving the solution synthesis of ZnMn-glycolate hollow microspheres and subsequent thermal annealing in air. When evaluated as an anode material for lithium-ion batteries, these ZnMn(2)O(4) ball-in-ball hollow microspheres
E P Leumann et al.
Pediatric nephrology (Berlin, Germany), 4(5), 493-497 (1990-09-01)
The molar ratios of oxalate and glycolate over creatinine were determined in single urine samples of 26 infants and 27 children aged 1-5 years. In 135 children aged 5-16 years, two urine specimens were collected, one before breakfast and one

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service