Skip to Content
Merck
All Photos(1)

Key Documents

92291

Supelco

Tricosafluorododecanoic acid

analytical standard

Synonym(s):

PFAS, Perfluorododecanoic acid, Perfluorolauric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)10CO2H
CAS Number:
Molecular Weight:
614.10
Beilstein:
1811257
EC Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for ISO 21675 2019

Assay

≥96.5% (GC)
96.5-103.0% (T)

shelf life

limited shelf life, expiry date on the label

bp

245 °C/740 mmHg (lit.)

mp

105-108 °C (lit.)

format

neat

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C12HF23O2/c13-2(14,1(36)37)3(15,16)4(17,18)5(19,20)6(21,22)7(23,24)8(25,26)9(27,28)10(29,30)11(31,32)12(33,34)35/h(H,36,37)

InChI key

CXGONMQFMIYUJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service