Skip to Content
Merck
All Photos(1)

Key Documents

91741

Sigma-Aldrich

Trimethylsilyl trifluoromethanesulfonate

purum, ≥98.0% (T)

Synonym(s):

TMS triflate, TMSOTf, Trifluoromethanesulfonic acid trimethylsilylester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Si(CH3)3
CAS Number:
Molecular Weight:
222.26
Beilstein:
1868911
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (T)

form

liquid

refractive index

n20/D 1.36 (lit.)

bp

77 °C/80 mmHg (lit.)

density

1.228 g/mL at 25 °C (lit.)

functional group

fluoro
triflate

SMILES string

C[Si](C)(C)OS(=O)(=O)C(F)(F)F

InChI

1S/C4H9F3O3SSi/c1-12(2,3)10-11(8,9)4(5,6)7/h1-3H3

InChI key

FTVLMFQEYACZNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethylsilyl trifluoromethanesulfonate may be used to catalyze:
  • Allylation of acetals to form homoallyl ethers.
  • Synthesis of 1,2-trans-glycosides.
  • Conversion of alcohols to esters.
  • Aminomethylation of silyl enol ethers with aminomethyl alkyl ethers.
  • Glycosidation of (+)-4-demethoxyanthracyclinones.

Other Notes

Efficient silylating agent and strong Lewis acid catalyst ; Review

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Bach, H. Brummerhop
J. Prakt. Chem., 341, 410-410 (1999)
Trimethylsilyl trifluoromethanesulfonate (trimethylsilyl triflate) as an excellent glycosidation reagent for anthracycline synthesis. Simple and efficient synthesis of optically pure 4-demethoxydaunorubicin.
Kimura Y
Chemistry Letters (Jpn), 13(4), 501-504 (1984)
An extremely fast and efficient acylation reaction of alcohols with acid anhydrides in the presence of trimethylsilyl trifluoromethanesulfonate as catalyst.
Procopiou PA
Chemical Communications (Cambridge, England), (23), 2625-2626 (1996)
Synthesis of homoallyl ethers via allylation of acetals in ionic liquids catalyzed by trimethylsilyl trifluoromethanesulfonate.
Zerth HM
Organic Letters, 5(1), 55-57 (2003)
Trimethylsilyl trifluoromethanesulfonate as an effective catalyst for glycoside synthesis.
Ogawa T
Carbohydrate Research, 93(1), C6-C9 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service