Skip to Content
Merck
All Photos(1)

Key Documents

91664

Supelco

Tridodecylmethylammonium nitrate

Selectophore, ≥99.0%

Synonym(s):

Methyltridodecylammonium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)11]3N(NO3)CH3
CAS Number:
Molecular Weight:
599.03
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

description

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥99.0% (T)
≥99.0%

form

powder

SMILES string

[O-][N+]([O-])=O.CCCCCCCCCCCC[N+](C)(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C37H78N.NO3/c1-5-8-11-14-17-20-23-26-29-32-35-38(4,36-33-30-27-24-21-18-15-12-9-6-2)37-34-31-28-25-22-19-16-13-10-7-3;2-1(3)4/h5-37H2,1-4H3;/q+1;-1

InChI key

AMIVMTKBZPOVKF-UHFFFAOYSA-N

General description

Tridodecylmethylammonium nitrate (TDMAN) is a nitrate ionophore which has good selectivity for nitrate more than chloride.
Visit our Sensor Applications portal to learn more.

Application

Selected application examples
TDMAN has been used in preparing liquid ion-selective electrode membrane during coulometric analysis of nitrate from fresh water samples.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Highly lipophilic electroactive component (nitrate ion-exchange) for solvent polymeric electrodes; used in ion-selective field effect transistors; Determination of nitrate in water, soil extracts and in agriculture by ion-selective electrode

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thin layer coulometric determination of nitrate in fresh waters.
Sohail M
Analytica Chimica Acta, 744, 39-44 (2012)
S. Wakida et al.
Bunseki Kogaku, 38, 510-510 (1989)
D. Wegmann et al.
Mikrochim. Acta III, 1-1 (1984)
V. Kratochvil
Ceskoslovenska Hygiena, 31, 355-355 (1986)
Hemin Wang et al.
Analytica chimica acta, 1129, 136-142 (2020-09-07)
Traditional potentiometric NO3--selective electrodes suffer from a fundamental limitation of the Nernst slope (59.1 mV/dec at 25 °C) due to the relationship between the potential and the logarithmic of ionic activity. Herein, a coulometric signal readout is proposed instead of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service