Skip to Content
Merck
All Photos(1)

Key Documents

91481

Supelco

Malathion

certified reference material, TraceCERT®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19O6PS2
CAS Number:
Molecular Weight:
330.36
Beilstein:
1804525
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

(storage under argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

shipped in

dry ice

storage temp.

-10 to -25°C

SMILES string

CCOC(=O)CC(SP(=S)(OC)OC)C(=O)OCC

InChI

1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3

InChI key

JXSJBGJIGXNWCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Maya L Groner et al.
Ecological applications : a publication of the Ecological Society of America, 23(6), 1443-1454 (2013-10-24)
For the past several decades, amphibian populations have been decreasing around the globe at an unprecedented rate. Batrachochytrium dendrobatidis (Bd), the fungal pathogen that causes chytridiomycosis in amphibians, is contributing to amphibian declines. Natural and anthropogenic environmental factors are hypothesized
Leslie C Alvarez et al.
Journal of medical entomology, 50(5), 1031-1039 (2013-11-05)
Resistance to the insecticides deltamethrin and malathion and the enzymes associated with metabolic resistance mechanisms were determined in four field populations of Aedes aegypti (L.) from western Venezuela during 2008 and 2010 using the bottle assay and the microplate biochemical
Guang-Mao Shen et al.
Pest management science, 68(12), 1553-1563 (2012-09-26)
The oriental fruit fly, Bactrocera dorsalis, is a major pest that infects fruits and agricultural products worldwide. The latest resistance monitoring of B. dorsalis from mainland China has identified high levels of resistance to insecticides. In this study, the biochemical
Hu Zhang et al.
Journal of agricultural and food chemistry, 60(41), 10188-10195 (2012-09-27)
An enantioselective method is presented for the determination of isocarbophos in soil by liquid chromatography coupled with tandem mass spectrometry. The pesticide residues in soil samples were extracted with acetonitrile, and complete enantioseparation was obtained on an amylose tris(3,5-dimethylphenylcarbamate) chiral
Mingjing Sun et al.
Journal of hazardous materials, 237-238, 140-146 (2012-09-12)
The dissipation behavior of the two enantiomers of malathion was elucidated in five plant species using enantioselective high performance liquid chromatography (HPLC), and the acute toxicity of the individual enantiomers toward earthworms and honeybees was studied. The calculated LC(50) values

Articles

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Chromatograms

application for LC-MS, application for SPE

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service