Skip to Content
Merck
All Photos(4)

Key Documents

88580

Sigma-Aldrich

Phenothiazine

purum, ≥98.0% (GC)

Synonym(s):

PTZ, 10H-Phenothiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9NS
CAS Number:
Molecular Weight:
199.27
Beilstein:
143237
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥98.0% (GC)
bp:
371 °C (lit.)

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

pellets

bp

371 °C (lit.)

mp

182-187 °C (lit.)
183-187 °C

SMILES string

N1c2ccccc2Sc3ccccc13

InChI

1S/C12H9NS/c1-3-7-11-9(5-1)13-10-6-2-4-8-12(10)14-11/h1-8,13H

InChI key

WJFKNYWRSNBZNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The structure of phenothiazine is rigid, being tricyclic. It is known to alter dopamine (3,4-dihydroxyphenethylamine). Its use as an electron donor is based on its unique hole transporting ability, electron releasing nitrogen and sulfur heteroatoms and its non-planar structure leading to lower molecular aggregation.

Application

Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photodegradation of trimeprazine triggered by self-photogenerated singlet molecular oxygen
Waseem A, et al
Journal of Saudi Chemical Society (2012)
Asif M, et al
Arabian Journal of Chemistry null
A S Horn et al.
Proceedings of the National Academy of Sciences of the United States of America, 68(10), 2325-2328 (1971-10-01)
Phenothiazines and butyrophenones are known to alter dopamine (3,4-dihydroxyphenethylamine) metabolism in the brain in a fashion suggesting that they may block dopamine receptors. We observed, using Dreiding molecular models, that dopamine in its solid-state conformation is superimposable upon a portion
Acridine and phenothiazine derivatives as pharmacotherapeutics for prion disease
Carsten K, et al
Proceedings of the National Academy of Sciences of the USA, 98(17), 9834-9841 (2001)
Mechanistic basis of phenothiazine-driven inhibition of Tau aggregation.
Elias Akoury et al.
Angewandte Chemie (International ed. in English), 52(12), 3511-3515 (2013-02-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service