87929
1-Hexyl-3-methylimidazolium chloride
≥97.0% (HPLC)
Synonym(s):
HMIMCl
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H19ClN2
CAS Number:
Molecular Weight:
202.72
Beilstein:
8356525
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥97.0% (HPLC)
form
liquid
SMILES string
[Cl-].CCCCCCn1cc[n+](C)c1
InChI
1S/C10H19N2.ClH/c1-3-4-5-6-7-12-9-8-11(2)10-12;/h8-10H,3-7H2,1-2H3;1H/q+1;/p-1
InChI key
NKRASMXHSQKLHA-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
1-hexyl-3-methylimidazolium chloride is an ionic liquid (IL). Its surface and bulk properties in aqueous solution at various temperatures indicates that it behaves as a short-chain cationic surfactant and shows aggregation behavior.
Physical form
Ionic solvent, liquid salt
Other Notes
Ionic liquid employed in the catalytic, solvent-free alkylation; catalyst for polymerization of olefins.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Effect of electrolytes on surface tension and surface adsorption of 1-hexyl-3-methylimidazolium chloride ionic liquid in aqueous solution.
Ghasemian E, et al.
The Journal of Chemical Thermodynamics, 42(8), 962-966 (2010)
Vasudevan V Namboodiri et al.
Chemical communications (Cambridge, England), (4), 342-343 (2002-07-18)
Microwave-assisted preparation of dialkylimidazolium tetrachloroaluminates, [CnMIM][AlCl4] and their application as recyclable catalysts for the efficient and eco-friendly protection of alcohols as tetrahydropyranyl (THP) ethers are described. The same catalyst can also be utilized for the deprotection of THP ethers.
Homogeneous Tritylation of Cellulose in 1?Butyl?3?methylimidazolium Chloride
Erdmenger, Tina, et al.
Macromolecular Bioscience, 7.4, 440-445 (2007)
Dynamic viscosities of a series of 1-alkyl-3-methylimidazolium chloride ionic liquids and their binary mixtures with water at several temperatures
Gomez, Elena, et al.
Journal of Chemical and Engineering Data, 51.2, 696-701 (2006)
Solution enthalpies of ionic liquid 1-hexyl-3-methylimidazolium chloride
Fang, Da-Wei, Ying-Chun Sun, and Zhen-Wei Wang
Journal of Chemical and Engineering Data, 53.1, 259-261 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service