Skip to Content
Merck
All Photos(2)

Key Documents

80342

Supelco

β-Boswellic acid

analytical standard

Synonym(s):

(3α,4β)-3-Hydroxyurs-12-en-23-oic acid, 3α-Hydroxyurs-12-en-24-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@@H]1CC[C@]2(C)CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@@H](O)[C@@](C)([C@@H]5CC[C@@]34C)C(O)=O)[C@@H]2[C@H]1C

InChI

1S/C30H48O3/c1-18-10-13-26(3)16-17-28(5)20(24(26)19(18)2)8-9-21-27(4)14-12-23(31)30(7,25(32)33)22(27)11-15-29(21,28)6/h8,18-19,21-24,31H,9-17H2,1-7H3,(H,32,33)/t18-,19+,21-,22-,23-,24+,26-,27-,28-,29-,30-/m1/s1

InChI key

NBGQZFQREPIKMG-PONOSELZSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

β-Boswellic acid is a naturally occurring pentacyclic triterpene, which can show pharmacological properties like antitumor, anticarcinogenic and antihyperlipidemic activity.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasrib Qurishi et al.
Future oncology (London, England), 8(7), 867-881 (2012-07-27)
Apoptotic induction in cancer cells has become a major focus of anticancer therapeutics. In this regard, β-boswellic acids, naturally occurring pentacyclic triterpenes, have demonstrated antiproliferative and cytotoxic effects against different types of cancers. Surprisingly, not much has been reported regarding
Acton A.Q et al.
Intracellular Membranes?Advances in Research and Application: 2013 Edition (2013)
Anti-tumor and anti-carcinogenic activities of triterpenoid,\beta-boswellic acid
Huang T-M, et al. et al.
Biofactors, 13, 225- 230 (2000)
Arne Henkel et al.
Biochemical pharmacology, 83(1), 115-121 (2011-10-18)
Lipophilic extracts of gum resins of Boswellia species (BSE) are used in folk medicine to treat various inflammatory disorders and infections. The molecular background of the beneficial pharmacological effects of such extracts is still unclear. Various boswellic acids (BAs) have
Sarita Saraswati et al.
Microvascular research, 82(3), 263-268 (2011-08-26)
The aim of the present study was to investigate the effects of boswellic acid (BA) on key components of inflammatory angiogenesis in the murine cannulated sponge implant angiogenesis model. Polyester-polyurethane sponges, used as a framework for fibrovascular tissue growth, were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service