Skip to Content
Merck
All Photos(5)

Documents

792535

Sigma-Aldrich

Malonic acid

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%

Synonym(s):

Propanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOH)2
CAS Number:
Molecular Weight:
104.06
Beilstein:
1751370
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

product line

ReagentPlus®
Redi-Dri

Assay

99%

form

powder

quality

free-flowing

mp

132-135 °C (dec.) (lit.)

solubility

1 M NaOH: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

OC(=O)CC(O)=O

InChI

1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)

InChI key

OFOBLEOULBTSOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • An eminent approach towards next generation solvents for sustainable packaging and stability of enzymes: a comprehensive study of ionic liquid and deep eutectic solvent mixtures.: This study explores the potential of malonic acid in forming deep eutectic solvents, which are utilized for enzyme stabilization and sustainable packaging solutions. The comprehensive analysis demonstrates the effectiveness of these solvents in various applications, highlighting their eco-friendly nature (Fatima et al., 2024).
  • Black Rice Anthocyanins as An Effective Antioxidant of Inhibition of Oil Oxidative Based on Molecular Modification.: This research investigates the use of malonic acid derivatives in modifying black rice anthocyanins to enhance their antioxidant properties. The findings suggest significant improvements in the stability and efficacy of these antioxidants in preventing oil oxidation (Zhao et al., 2024).
  • Determination of trace chelating carboxylic acids in rice by green extraction combined with liquid chromatography-mass spectrometry analysis and its application in the evaluation of old and new rice.: The application of malonic acid in this study focuses on its use in green extraction methods for determining trace amounts of chelating carboxylic acids in rice. This method shows promise in evaluating the quality and age of rice samples (Kang et al., 2024).
  • Molecular marker identification, antioxidant, antinociceptive, and anti-inflammatory responsiveness of malonic acid capped silver nanoparticle.: This research highlights the multifunctional applications of malonic acid-capped silver nanoparticles. These nanoparticles exhibit strong antioxidant, antinociceptive, and anti-inflammatory properties, making them useful in various biomedical applications (Fatima et al., 2023).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service