Skip to Content
Merck
All Photos(1)

Key Documents

78769

Supelco

NMP

analytical standard

Synonym(s):

1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein:
106420
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

1-Methyl-2-pyrrolidinone, analytical standard

grade

analytical standard

Quality Level

vapor density

3.4 (vs air)

vapor pressure

0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

Assay

≥99.9% (GC)

autoignition temp.

518 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

9.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.47 (lit.)
n20/D 1.470

bp

202 °C (lit.)
81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CN1CCCC1=O

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Methyl-2-pyrrolidinone is a polar solvent widely used in organic chemistry and polymer chemistry. On a large scale, it finds applications in the recovery and purification of acetylenes, olefins, and diolefins, gas purification, and aromatics extraction from feedstocks.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

1-Methyl-2-pyrrolidinone may be used as an eluent and extraction solvent for the extraction of soluble materials in soil samples using size exclusion chromatography.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xingxing Gu et al.
Frontiers in chemistry, 8, 149-149 (2020-03-21)
A Typha Angustifolia-like MoS2/carbon nanofiber composite as both a chemically trapping agent and redox conversion catalyst for lithium polysulfides has been successfully synthesized via a simple hydrothermal method. Cycling performance and coulombic efficiency have been improved significantly by applying the
B A Jönsson et al.
Journal of chromatography. B, Biomedical sciences and applications, 694(2), 351-357 (1997-07-04)
A method for simultaneous determination of 5-hydroxy-N-methylpyrrolidone and 2-hydroxy-N-methylsuccinimide in urine is described. These compounds are metabolites of N-methyl-2-pyrrolidone, a powerful and widely used organic solvent. 5-Hydroxy-N-methylpyrrolidone and 2-hydroxy-N-methylsuccinimide were purified from urine by adsorption to a C8 solid-phase extraction
Chun-Wa Chung et al.
Journal of medicinal chemistry, 55(2), 576-586 (2011-12-06)
Bromodomain-containing proteins are key epigenetic regulators of gene transcription and readers of the histone code. However, the therapeutic benefits of modulating this target class are largely unexplored due to the lack of suitable chemical probes. This article describes the generation
David S Hewings et al.
Journal of medicinal chemistry, 54(19), 6761-6770 (2011-08-20)
Histone-lysine acetylation is a vital chromatin post-translational modification involved in the epigenetic regulation of gene transcription. Bromodomains bind acetylated lysines, acting as readers of the histone-acetylation code. Competitive inhibitors of this interaction have antiproliferative and anti-inflammatory properties. With 57 distinct
Ajit Dhananjay Jagtap et al.
European journal of medicinal chemistry, 85, 268-288 (2014-08-05)
A series of 6-acylureido derivatives containing a 3-(pyrrol-2-ylmethylidene)indolin-2-one scaffold were synthesized as potential dual Aurora B/FLT3 inhibitors by replacing the 6-arylureido moiety in 6-arylureidoindolin-2-one-based multi-kinase inhibitors. (Z)-N-(2-(pyrrolidin-1-yl)ethyl)-5-((6-(3-(2-fluoro-4-methoxybenzoyl)ureido)-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide (54) was identified as a dual Aurora B/FLT3 inhibitor (IC50 = 0.4 nM and 0.5 nM, respectively).

Articles

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Questions

  1. Is the unit for the purity (GC) in %w/w or on %area

    1 answer
    1. The purity by GC is in area percentage.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service