Skip to Content
Merck
All Photos(1)

Key Documents

77947

Supelco

4-(2-Hydroxy-3-isopropylaminopropoxy)phenylacetic acid

analytical standard

Synonym(s):

4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetic acid, Atenolol acid, Metoprolol acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21NO4
CAS Number:
Molecular Weight:
267.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)NCC(O)COc1ccc(CC(O)=O)cc1

InChI

1S/C14H21NO4/c1-10(2)15-8-12(16)9-19-13-5-3-11(4-6-13)7-14(17)18/h3-6,10,12,15-16H,7-9H2,1-2H3,(H,17,18)

InChI key

PUQIRTNPJRFRCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(2-Hydroxy-3-isopropylaminopropoxy)phenylacetic acid is a metabolite of the selective β-1 blocker drug metoprolol.

Application

4-(2-Hydroxy-3-isopropylaminopropoxy)phenylacetic acid may be used as an analytical standard for the determination of the analyte in environmental water samples, wastewater samples, and human and animal biological samples by various chromatography techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The
Small Animal Clinical Pharmacology, 241, 484-493 (2011)
Investigation of pharmaceuticals and their metabolites in Brazilian hospital wastewater by LC-QTOF MS screening combined with a preliminary exposure and in silico risk assessment
Becker RW, et al.
The Science of the Total Environment, 699, 134218-134218 (2020)
Screening hundreds of emerging organic pollutants (EOPs) in surface water from the Yangtze River Delta (YRD): occurrence, distribution, ecological risk
Peng Y, et al.
Environmental Pollution (Barking, Essex : 1987), 241, 484-493 (2018)
Evaluation of in-situ calibration of Chemcatcher passive samplers for 322 micropollutants in agricultural and urban affected rivers
Moschet C, et al.
Water Research, 71, 306-317 (2015)
Determination of metoprolol, and its four metabolites in dog plasma
Fang J, et al.
Journal of Chromatography. B, Biomedical Applications, 809(1), 9-14 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service