Skip to Content
Merck
All Photos(1)

Key Documents

76778

Supelco

Methyl acrylate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOOCH3
CAS Number:
Molecular Weight:
86.09
Beilstein:
605396
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

3 (vs air)

vapor pressure

67.5 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

874 °F

shelf life

limited shelf life, expiry date on the label

contains

~0.0015% hydroquinone monomethyl ether as stabilizer

expl. lim.

14.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.402 (lit.)
n20/D 1.403

bp

80 °C (lit.)

mp

−75 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

SMILES string

COC(=O)C=C

InChI

1S/C4H6O2/c1-3-4(5)6-2/h3H,1H2,2H3

InChI key

BAPJBEWLBFYGME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Methyl acrylate can undergo transesterification reaction with 1-butanol under thermal conditions in the presence of hydrochloric acid to yield butyl acrylate as the major product.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

27.0 °F - closed cup

Flash Point(C)

-2.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Chemistry (2008)
Jens Voepel et al.
Biomacromolecules, 12(1), 253-259 (2010-12-21)
A multifunctional macroinitiator for single-electron-transfer mediated living radical polymerization (SET-LRP) was designed from acetylated galactoglucomannan (AcGGM) by α-bromoisobutyric acid functionalization of the anomeric hydroxyl groups on the heteropolysaccharide backbone. This macroinitiator, with a degree of substitution of 0.15, was used
Mustafa Ciftci et al.
Chemical communications (Cambridge, England), 48(82), 10252-10254 (2012-09-13)
Lightly branched, hyperbranched and cross-linked polymers with clickable sites were synthesized via a modified version of self-condensing photoinitiated copolymerization of methyl acrylate (MA) with propargyl acrylate (PA). The method is based on the use of a PA monomer containing two
Daniel Duncan et al.
Dalton transactions (Cambridge, England : 2003), 40(9), 1998-2005 (2011-01-26)
A new fluorous PCP pincer ligand has been coordinated to Ni(II), Pd(II) and Pt(II). The air stable palladium complex, which promotes Heck reactions between methyl acrylate and either aryl bromides or iodides, can be recovered intact by fluorous solid-phase extraction
Cláudio G L Junior et al.
Bioorganic chemistry, 38(6), 279-284 (2010-09-22)
Sixteen aromatic Morita-Baylis-Hillman adducts (MBHA) 1-16 were efficiently synthesized in a one step Morita-Baylis-Hillman reaction (MBHR) involving commercial aldehydes with methyl acrylate or acrylonitrile (81-100% yields) without the formation of side products on DABCO catalysis and at low temperature (0°C).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service