Skip to Content
Merck
All Photos(2)

Documents

71734

Supelco

Sodium ionophore III

Selectophore, function tested

Synonym(s):

ETH 2120, N,N,N′,N′-Tetracyclohexyl-1,2-phenylenedioxydiacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H52N2O4
CAS Number:
Molecular Weight:
552.79
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥98.0% (HPLC)

form

crystals

quality

function tested

SMILES string

O=C(COc1ccccc1OCC(=O)N(C2CCCCC2)C3CCCCC3)N(C4CCCCC4)C5CCCCC5

InChI

1S/C34H52N2O4/c37-33(35(27-15-5-1-6-16-27)28-17-7-2-8-18-28)25-39-31-23-13-14-24-32(31)40-26-34(38)36(29-19-9-3-10-20-29)30-21-11-4-12-22-30/h13-14,23-24,27-30H,1-12,15-22,25-26H2

InChI key

GKRBLFCTFPAHMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium ionophore III is generally used for preparing ion-selective electrode.
Visit our Sensor Applications portal to learn more.

Application

Selected application examples
It may be used in the preparation of vulcanizing-type silicone rubber-based sodium-selective membrane electrode.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ionophore suitable for the assay of Na+-activity in blood, plasma, serum. etc. with a solvent polymeric membrane electrode

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T. Maruizumi et al.
Mikrochimica Acta, I, 331-331 (1986)
Zachary J Sandler et al.
Antimicrobial agents and chemotherapy, 64(6) (2020-04-15)
Bunyaviruses are significant human pathogens, causing diseases ranging from hemorrhagic fevers to encephalitis. Among these viruses, La Crosse virus (LACV), a member of the California serogroup, circulates in the eastern and midwestern United States. While LACV infection is often asymptomatic
One-component room temperature vulcanizing-type silicone rubber-based sodium-selective membrane electrodes.
Poplawski, Michael E., et al.
Analytica Chimica Acta, 355.2, 249-257 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service