Skip to Content
Merck
All Photos(1)

Key Documents

70191

Millipore

Mueller Hinton Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

MH agar; Antibiotic sensitivity test media; disc diffusion test media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 17.0 g/L
beef infusion solids, 2.0 g/L
casein hydrolysate, 17.5 g/L
starch, 1.5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | susceptibility testing: suitable
microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

application(s)

clinical testing
food and beverages

microbiology

storage temp.

10-30°C

suitability

nonselective for Neisseria spp.
nonselective for bacteria (General Media)

General description

Mueller Hinton Agar is specially designed for antibiotic susceptibility testing using the disc diffusion method (Kirby Bauer method). It has been recommended by the CLSI as the ideal medium for antibiotic susceptibility testing for the following reasons:

  • It shows good reproducibility from batch to batch
  • It is low in certain inhibitors like trimethoprim, sulfonamide and tetracycline
  • It supports the growth of both fastidious and non-fastidious bacterial pathogens
  • The starch in the media absorbs toxins thus preventing them from interfering with the antibiotics
  • It is a loose agar and therefore facilitates better diffusion of antibiotics
Mueller Hinton Agar has been used:
  • In culturing Escherichia coli ATCC™ 25922 strain for disc diffusion studies
  • In disk diffusion assay of fusidic acid against Staphylococcus aureus, Bacillus cereus, Pseudomonas aeruginosa and Escherichia coli
  • In agar diffusion assay of drug-loaded nano-hydroxyapatite formulations against Staphylococcus aureus, Pseudomonas aeruginosa and Escherichia coli

Application

Mueller Hinton Agar is a solid medium originally designed for the isolation of pathogenic Neisseria species, now widely used for antibiotic susceptibility testing (including sulfonamides) of aerobic and facultatively anaerobic bacteria isolated from clinical specimens.

Preparation Note

Suspend 38 g in 1 litre of distilled water, bring to the boil to dissolve the medium completely and sterilize by autoclaving at 121°C for 15 minutes.

Other Notes

Enhancement of cefotoxamine and other cephalosporins against Enterococcus faecalis by blood supplemented Mueller-Hinton agar

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multi-drug delivery system based on lipid membrane mimetic coated nano-hydroxyapatite formulations
Placente D, et al.
International Journal of Pharmaceutics, 548(1), 559-570 (2018)
Preparation and evaluation of QbD based fusidic acid loaded in situ gel formulations for burn wound treatment
Aksu NB, et al.
Journal of drug delivery science and technology, 52, 110-121 (2019)
Solid-state structure and antimicrobial and cytotoxicity studies of a cucurbit [6] uril-like Cu6L4 constructed from 3, 5-bis [(1H-tetrazol-5-yl) methyl]-4H-1, 2, 4-triazol-4-amine
Vasylevskyi S, et al.
Acta Crystallographica Section C, Structural Chemistry, 74(11) (2018)
R D Jenkins et al.
Journal of clinical microbiology, 22(3), 369-374 (1985-09-01)
Disk diffusion susceptibility tests for enterococci are frequently modified by adding 5% sheep blood (SB) to Mueller-Hinton agar; the performance standards from the National Committee for Clinical Laboratory Standards sanction this addition. Susceptibility testing of aminoglycoside antibiotics is not recommended
Luciano Benedini et al.
Materials science & engineering. C, Materials for biological applications, 115, 111101-111101 (2020-07-01)
Bone substitute materials based on bioceramics and polymers have evolved shifting from a passive role where they are merely accepted by the body; to an active role, where they respond to particular environmental conditions or to different types of cues

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service