Skip to Content
Merck
All Photos(1)

Documents

68186

Sigma-Aldrich

Benzylpenicilloic acid disodium salt, mixture of epimeres

≥95% (HPLC)

Synonym(s):

4-Carboxy-5,5-dimethyl-α-[(2-phenylacetyl)amino]-2-thiazolidineacetic acid disodium salt, Penicilloic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N2Na2O5S
CAS Number:
Molecular Weight:
396.37
UNSPSC Code:
12352106
NACRES:
NA.85

Quality Level

Assay

≥95% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

format

neat

Mode of action

cell wall synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

InChI

1S/C16H20N2O5S/c1-16(2)12(15(22)23)18-13(24-16)11(14(20)21)17-10(19)8-9-6-4-3-5-7-9/h3-7,11-13,18H,8H2,1-2H3,(H,17,19)(H,20,21)(H,22,23)

InChI key

HCYWNSXLUZRKJX-UHFFFAOYSA-N

General description

This product is a penicilloic acid with the N-acyl group phenylacetyl as side-chain. In alkaline medium, 5R,6R-Benzylpenicilloic acid epimerizes slowly to 5S,6R-benzylpenicilloic acid until equilibrium.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuangji Liu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(7-8), 533-540 (2011-02-09)
A rapid, sensitive, and specific method for the determination of amoxicillin (AMO), amoxicilloic acid (AMA), amoxicillin diketopiperazine-2',5'-dione (DIKETO), penicillin G (PEN G), benzylpenicilloic acid (BPA-1), benzylpenilloic acid (BPA-2), and benzylpenillic acid (BPA-3) in bovine milk using ultra-high-performance liquid chromatography-tandem mass
Sander S van Berkel et al.
ACS chemical biology, 8(10), 2112-2116 (2013-08-01)
β-Lactam antibiotics react with penicillin binding proteins (PBPs) to form relatively stable acyl-enzyme complexes. We describe structures derived from the reaction of piperacillin with PBP3 (Pseudomonas aeruginosa) including not only the anticipated acyl-enzyme complex but also an unprecedented complex with
Reza M Seifar et al.
Journal of chromatography. A, 1187(1-2), 103-110 (2008-02-26)
A rapid, sensitive and selective ion-pair reversed-phase liquid chromatography-electrospray ionization isotope dilution tandem mass spectrometry (IP-LC-ESI-ID-MS/MS) was developed for quantitative analysis of free intracellular metabolites in cell cultures. As an application a group of compounds involved in penicillin biosynthesis pathway
I Ghebre-Sellassie et al.
Journal of pharmaceutical sciences, 73(1), 125-128 (1984-01-01)
5R,6R-Benzylpenicilloic acid was found to epimerize slowly in alkaline media to 5S,6R-benzylpenicilloic acid until equilibrium was established. Epimerization proceeded via the imine tautomer of penamaldic acid rather than the enamine form and was found to favor the 5S,6R-epimer at equilibrium.
Degradation of benzylpenicillin at pH 7.5 to D-benzylpenicilloic acid.
B B LEVINE
Nature, 187, 939-940 (1960-09-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service