Skip to Content
Merck
All Photos(4)

Key Documents

675970

Sigma-Aldrich

Cyclopentyl methyl ether

greener alternative

contains 50 ppm BHT as inhibitor, anhydrous, ≥99.9%

Synonym(s):

CPME

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O
CAS Number:
Molecular Weight:
100.16
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
106 °C/760 mmHg
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Assay

≥99.9%

form

liquid

contains

50 ppm BHT as inhibitor

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.0050% H2O
≤0.0050% water
water

≤50 ppm H2O2

evapn. residue

<0.003%

bp

106 °C/760 mmHg

mp

-140 °C (lit.)

density

0.86 g/mL at 25 °C

greener alternative category

SMILES string

COC1CCCC1

InChI

1S/C6H12O/c1-7-6-4-2-3-5-6/h6H,2-5H2,1H3

InChI key

SKTCDJAMAYNROS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Cyclopentyl methyl ether (CPME) is a greener alternative to other ethereal solvents such as tetrahydrofuran (THF), 2-methyl tetrahydrofuran (2-MeTHF), dioxane (carcinogenic), tert-Butyl methyl ether (MTBE), and 1,2-dimethoxyethane (DME) and thus has been enhanced for "Safer Solvents and Auxiliaries". Click here for more information.

Application

  • Cyclopentyl methyl ether (CPME) is an alternative to ether solvents such as THF, diethyl ether, and MTBE with a higher resistance to peroxide formation.[1]
  • The more environmentally conservative CPME solvent replaces hazardous solvents in order to achieve sustainability and reduce environmental and operational costs.[2]
  • CPME offers considerable potential and advantage as a direct replacement for CH2Cl2 in binary eluents using MeOH as the modifier with MeOH–DMC and i-PrOH–EtOAc.[2]
Green Alternative to THF, tert-Butyl methyl ether (MTBE), 1,4 Dioxane and other ether solvents

The toxicological assessment of cyclopentyl methyl ether (CPME) as a green solvent.

Features and Benefits

Greener alternative for THF, Diethyl ether, and MTBE

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

<37.4 °F - closed cup

Flash Point(C)

< 3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclopentyl Methyl Ether as a New and Alternative Process Solvent
Watanabe, K.; Yamagiwa, N.; Torisawa, Y.
Organic Process Research & Development, 11 (2007)
Replacement of dichloromethane within chromatographic purification: a guide to alternative solvents
MacMillan, D. S.; et al.
Green Chemistry (2012)
Rikako Takahiro et al.
Biological & pharmaceutical bulletin, 38(1), 58-65 (2015-03-07)
The aim of this study was to characterize the kinetics of metabolite formation of the phosphodiesterase type-5 (PDE5) inhibitors sildenafil and tadalafil by CYP3A4, CYP3A5, and CYP3A7 isoforms. The formations of N-desmethyl sildenafil and desmethylene tadalafil were examined using CYP3A

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service