Skip to Content
Merck
All Photos(1)

Documents

67262

Supelco

2-Ethylhexyl acrylate

analytical standard

Synonym(s):

(±)-Acrylic acid 2-ethylhexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOOCH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
184.28
Beilstein:
1765828
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

6.4 (vs air)

vapor pressure

0.15 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

496 °F

shelf life

limited shelf life, expiry date on the label

contains

0.001-0.11% monomethyl ether hydroquinone as stabilizer

expl. lim.

6.4 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.436 (lit.)
n20/D 1.436

bp

215-219 °C (lit.)

density

0.884 g/mL at 20 °C
0.885 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCC(CC)COC(=O)C=C

InChI

1S/C11H20O2/c1-4-7-8-10(5-2)9-13-11(12)6-3/h6,10H,3-5,7-9H2,1-2H3

InChI key

GOXQRTZXKQZDDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylhexyl acrylates are classified under the ester group of compounds.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

2-Ethylhexyl acrylate has been used as a reference standard for the analysis of 2-ethylhexyl acrylate in glossy water-based lacquer by thermal desorption and gas chromatography-mass spectrometry (GC-MS).
It may be used as a reference standard for the determination of 2-ethylhexyl acrylate in:
  • Exhaled breath samples from cigarette smokers by capillary micro-extracton (CMV) coupled with GC-MS.
  • Food paper contact papers by modified quick, easy, cheap, effective, rugged and safe (QuEchERS) extraction procedure combined with gas chromatography-tandem mass spectrometry (GC-MS/MS).
  • Polymer emulsions by headspace (HS) GC-MS operating on selected ion monitoring (SIM) mode.
  • Adhesives employed in food contact materials by ultra-performance liquid chromatography-quadrupole time-of-flight mass spectrometry (UPLC-QTOF/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of a novel reference material in an international round robin test on material emissions testing.
Horn W, et al.
Indoor Air, 28(1), 181-187 (2018)
Risk assessment derived from migrants identified in several adhesives commonly used in food contact materials
Canellas E, et al.
Food And Chemical Toxicology, 75(19), 79-87 (2015)
Determination of residual (free) monomer in water based polymer emulsions by head space gas chromatography mass spectrometry.
Petha NH, et al.
Polymer Testing, 63(19), 462-466 (2017)
Analysis of exhaled breath from cigarette smokers using CMV-GC/MS.
Hamblin DN and Almirall J
Forensic Chemistry, 4, 67-74 (2017)
Simultaneous determination of seven acrylates in food contact paper products by GC/MS and modified QuEChERS.
Xun Z, et al.
Analytical Methods : Advancing Methods and Applications, 8(19), 3953-3958 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service