Skip to Content
Merck
All Photos(1)

Key Documents

63016

Millipore

M-17 Agar

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12.75 g/L
ascorbic acid, 0.5 g/L
casein peptone (tryptic), 2.5 g/L
disodium β-glycerophosphate pentahydrate, 19 g/L
magnesium sulfate hydrate, 0.25 g/L
meat extract, 5 g/L
meat peptone (peptic), 2.5 g/L
soyapeptone (papainic), 5 g/L
yeast extract, 2.5 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

7.1±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages

microbiology

suitability

nonselective for Lactococcus spp.
nonselective for Streptococcus spp.

Application

M-17 Agar media is proposed for the cultivation, detection and enumeration of Iactic streptococci in milk and dairy products. The M-17 media are superior to other comparable culture media for the cultivation of the fastidious species Streptococcus cremoris, Streptococcus diacetilactis and Streptococcus Iactis. It is also suitable for cultivation and maintenance of starter cultures for cheese and yogurt manufacturing. This medium helps in detecting streptococcal mutants which are lactose non-fermenters. M-17 Agar media may also be used for the differentiation of bacteriophages infecting lactic streptococci.

Preparation Note

Dissolve 50 g in 950 ml distilled water and sterilize by autoclaving at 121°C for 15 minutes. After cooling add 50 ml aseptic 10% lactose solution.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B P Keogh
Applied and environmental microbiology, 40(4), 798-802 (1980-10-01)
To assess the relative merits of tryptone yeast extract agar, the same medium unbuffered, and medium M17 for the assay of nine bacteriophages of lactic streptococci, comparative plaque counts were made with an overlay of 3 or 9 ml. Four
John Samelis et al.
Food microbiology, 28(1), 76-83 (2010-11-09)
This study used a combination of phenotypic, physical (Fourier Transformed Infra-Red [FTIR] spectroscopy) and molecular (RFLP and SSCP analysis of 16S rRNA genes) methods to identify the lactic acid bacteria (LAB) flora present in traditional Greek Graviera cheese after five
Natan Gollop et al.
Biotechnology progress, 19(2), 436-439 (2003-04-05)
Propionibacterium thoenii P-127 produces and releases to the growth medium antibacterial agents that can be used as natural preservatives. The concentrations of these antibacterial agents in the growth medium are very low, and their activity can be detected only in
Eleftheria Barouni et al.
Food chemistry, 200, 76-82 (2016-02-03)
The object of the current study was to develop a new process for continuous Feta-type cheese production using a biocatalyst consisting of immobilized rennin on a tubular cellulose/starch gel (TC/SG) composite, which has been proven to be an appropriate carrier

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service