Skip to Content
Merck
All Photos(1)

Key Documents

53008

Supelco

Diethyl phthalate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C2H5)2
CAS Number:
Molecular Weight:
222.24
Beilstein:
1912500
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

7.66 (vs air)

vapor pressure

1 mmHg ( 100 °C)

product line

PESTANAL®

Assay

≥99.5% (GC)

autoignition temp.

854 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

0.75 %, 187 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.502 (lit.)
n20/D 1.502

bp

298-299 °C (lit.)

mp

−3 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCOC(=O)c1ccccc1C(=O)OCC

InChI

1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3

InChI key

FLKPEMZONWLCSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl phthalate belongs to the class of phthalates, widely used as solvents and odorless diluents in perfumes and cosmetics.[1]

Application

Diethyl phthalate is used as an analytical reference standard for the quantification of the analyte in cosmetics,[1] water and beverages[2] using different chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction of phthalate esters from water and beverages using a graphene-based magnetic nanocomposite prior to their determination by HPLC
Wu Q, et al.
Microchimica Acta, 177(1-2), 23-30 (2012)
Simultaneous determination of seven phthalates and four parabens in cosmetic products using HPLC-DAD and GC-MS methods
Shen Y-H, et al.
Journal of Separation Science, 30(1), 48-54 (2007)
Lee-Ju Cheng et al.
Aquatic toxicology (Amsterdam, Netherlands), 109, 166-175 (2011-10-28)
The toxicity and effects of diethyl phthalate (DEP), a potent allelochemical, on the growth of greater duckweed were studied. Biochemical analyses and physiological methods were combined to investigate oxidative stress, adverse effects and their mechanisms in greater duckweeds grown in
Maik Hadorn et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(50), 20320-20325 (2012-11-24)
Higher-order structures that originate from the specific and reversible DNA-directed self-assembly of microscopic building blocks hold great promise for future technologies. Here, we functionalized biotinylated soft colloid oil-in-water emulsion droplets with biotinylated single-stranded DNA oligonucleotides using streptavidin as an intermediary
Antonia M Calafat et al.
Environmental health perspectives, 114(11), 1783-1789 (2006-11-17)
The probability of nonoccupational exposure to phthalates is high given their use in a vast range of consumables, including personal care products (e.g., perfumes, lotions, cosmetics), paints, industrial plastics, and certain medical devices and pharmaceuticals. Phthalates are of high interest

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service