Skip to Content
Merck
All Photos(3)

Key Documents

484407

Sigma-Aldrich

1-Methoxy-2-propanol

greener alternative

≥99.5%

Synonym(s):

Propylene glycol methyl ether, Propyleneglycol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OCH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
1731270
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
application(s):
microbiology
bp:
118-119 °C (lit.)
vapor pressure:
10.9 mmHg ( 25 °C)
Pricing and availability is not currently available.

vapor density

3.12 (vs air)

Quality Level

vapor pressure

10.9 mmHg ( 25 °C)

product line

ReagentPlus®

Assay

≥99.5%

form

liquid

autoignition temp.

532 °F

expl. lim.

13.8 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.001% water

refractive index

n20/D 1.403 (lit.)

bp

118-119 °C (lit.)

mp

-97 °C

solubility

water: miscible

density

0.916 g/mL at 25 °C (lit.)

application(s)

microbiology

greener alternative category

SMILES string

CC(O)COC

InChI

1S/C4H10O2/c1-4(5)3-6-2/h4-5H,3H2,1-2H3

InChI key

ARXJGSRGQADJSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Methoxy-2-propanol (Propylene glycol methyl ether, PGME), a glycol ether, can be synthesized by reacting propylene oxide with methanol in the presence of ZnMgAl (zinc-magnesium-aluminium) catalysts.[1] Its degradation by microorganisms in different soil types has been investigated.[2] An oral reference dose (RfD) and an inhalation reference concentration (RfC) values of PGME have been obtained from inhalation studies in F344 rats and B6C3F1 mice.[3]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry.Monoethers, especially propylene glycol methyl ether (PGME), are widely used as pollution-free solvents in fine chemical synthesis in pharmaceutical, chemical and cosmetic industries and thus aligns with "Safer Solvents and Auxiliaries". Click here for more information.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

88.7 °F - closed cup - (External MSDS)

Flash Point(C)

31.5 °C - closed cup - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Influence of acid-base properties of ZnMgAl-mixed oxides for the synthesis of 1-methoxy-2-propanol.
Cheng W, et al.
Applied Clay Science, 42(1), 111-115 (2008)
Biodegradation of glycol ethers in soil.
Gonsior SJ and West RJ.
Environmental Toxicology and Chemistry / Setac, 14(8), 1273-1279 (1995)
Rei Momose et al.
Organic letters, 15(8), 2010-2013 (2013-04-11)
Three new alkaloids, hyrtimomines A-C (1-3), were isolated from an Okinawan marine sponge Hyrtios sp. The structures of 1-3 were elucidated on the basis of spectroscopic analysis and application of a phenylglycine methyl ester (PGME) method. Hyrtimomines A (1) and
W R Berendsen et al.
Biotechnology and bioengineering, 95(5), 883-892 (2006-08-29)
Lipase-catalyzed kinetic resolution of racemates is a popular method for synthesis of chiral synthons. Most of these resolutions are reversible equilibrium limited reactions. For the first time, an extensive kinetic model is proposed for kinetic resolution reactions, which takes into
S R Vink et al.
Regulatory toxicology and pharmacology : RTP, 58(1), 64-71 (2010-04-17)
REACH requests the exploration of alternative strategies for hazard identification before resorting to (in vivo) testing. Here, we combined read-across as non-testing strategy with a tiered exposure assessment for the risk characterisation of 1-methoxypropan-2-ol (PGME) as a representative for phase-in

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service