Skip to Content
Merck
All Photos(1)

Key Documents

46959

Supelco

Tiamulin fumarate

VETRANAL®, analytical standard

Synonym(s):

Thiamutilin fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H51NO8S
CAS Number:
Molecular Weight:
609.81
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\C(O)=O.CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)[C@@]23CC[C@@H](C)[C@]1(C)[C@@H]2C(=O)CC3

InChI

1S/C28H47NO4S.C4H4O4/c1-8-26(6)17-22(33-23(31)18-34-16-15-29(9-2)10-3)27(7)19(4)11-13-28(20(5)25(26)32)14-12-21(30)24(27)28;5-3(6)1-2-4(7)8/h8,19-20,22,24-25,32H,1,9-18H2,2-7H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/t19?,20-,22+,24-,25-,26+,27-,28-;/m0./s1

InChI key

YXQXDXAHCSEVSD-IHAONTCTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sigma-Aldrich offers tiamulin fumarate for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyses. Purity of the chemical is about 99% unless otherwise stated.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petra Reinhold et al.
Veterinary journal (London, England : 1997), 187(3), 405-407 (2010-08-31)
The effect of short-term antimicrobial treatment on natural excretion of Chlamydia suis in rectal swabs and C. suis and Chlamydophila psittaci in nasal swabs was investigated in 47 clinically normal piglets by quantitative real-time PCR. Pigs were treated IM with
K M S Islam et al.
Poultry science, 87(12), 2528-2534 (2008-11-29)
In an earlier study, the continuous medication of broiler feed with a combination of tiamulin (TIA; 20 mg/kg), chlortetracycline (CTC; 60 mg/kg), and the ionophore anticoccidial salinomycin (SAL; 60 mg/kg) caused an initial increase in BW and feed efficiency (FE;
Xun Pan et al.
Journal of environmental sciences (China), 23(10), 1729-1737 (2011-01-01)
This work describes a systematic approach to the development of a method for simultaneous determination of three classes of veterinary antibiotics in the suspended solids (SS) of swine wastewater, including five sulfonamides, three tetracyclines and one macrolide (tiamulin). The entire
Saad Gharaibeh et al.
Veterinary microbiology, 150(3-4), 379-383 (2011-03-09)
This study compares the antimicrobial susceptibility over time between two groups of Mycoplasma gallisepticum (MG) isolates from the same geographical area. Minimum inhibitory concentration of 13 antimicrobials was determined against two groups of MG isolates from chickens. Group 1 strains
Line Lolk et al.
Journal of medicinal chemistry, 51(16), 4957-4967 (2008-08-06)
Pleuromutilin and its derivatives are antibacterial drugs that inhibit protein synthesis in bacteria by binding to ribosomes. To promote rational design of pleuromutilin based drugs, 19 pleuromutilin conjugates with different nucleoside fragments as side chain extensions were synthesized by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service