Skip to Content
Merck
All Photos(1)

Documents

46874

Supelco

Sulfanilamide

VETRANAL®, analytical standard

Synonym(s):

p-Aminobenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4SO2NH2
CAS Number:
Molecular Weight:
172.20
Beilstein:
511852
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39093202
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

164-166 °C (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

clinical testing

format

neat

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

Nc1ccc(cc1)S(N)(=O)=O

InChI

1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)

InChI key

FDDDEECHVMSUSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide
Sulfanilamide is an antibacterial drug.

Application

It was used in a study to demonstrate photodecomposition in aqueous solution of cutaneous photosensitizing agents with the help of spin traps 5, 5-dimethyl-1-pyrroline-1-oxide.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Claudia Temperini et al.
Journal of medicinal chemistry, 53(2), 850-854 (2009-12-24)
Coumarins constitute a general and totally new class of inhibitors of the zinc enzyme carbonic anhydrase (CA, EC 4.2.1.1), binding at the entrance of the active site cavity. We report here that the coumarin-binding site in CAs may interact with
SPECTROSCOPIC STUDIES OF CUTANEOUS PHOTOSENSITIZING AGENTS?II. SPIN TRAPPING OF PHOTOLYSIS PRODUCTS FROM SULFANILAMIDE AND 4-AMINOBENZOIC ACID USING 5, 5-DIMETHYL-1-PYRROLINE-1-OXIDE.
Chignell, Colin F., et al.
Photochemistry and Photobiology, 34.2 , 147-156 (1981)
Edward E Knaus et al.
Bioorganic & medicinal chemistry letters, 21(19), 5892-5896 (2011-08-20)
A series of compounds incorporating regioisomeric phenylethynylbenzenesulfonamide moieties has been investigated for the inhibition of four human carbonic anhydrase (hCA, EC 4.2.1.1) isoforms, hCA I, II, IX and XII. Inhibition between the low nanomolar to the milliomolar range has been
Penghua Wang et al.
Water research, 45(16), 5015-5026 (2011-08-02)
A novel carbon-sensitized and nitrogen-doped TiO2 (C/N-TiO2) was synthesized by a facile sol-gel method using titanium butoxide as both titanium precursor and carbon source, and nitric acid as nitrogen source. The calcination temperature had a great effect on the crystal
Electrophilic trifluoromethanesulfanylation of organometallic species with trifluoromethanesulfanamides.
François Baert et al.
Angewandte Chemie (International ed. in English), 51(41), 10382-10385 (2012-09-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service