Skip to Content
Merck
All Photos(1)

Key Documents

46434

Supelco

3-Methylcholanthrene solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16
CAS Number:
Molecular Weight:
268.35
Beilstein:
1913890
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Cc1ccc2cc3c(ccc4ccccc34)c5CCc1c25

InChI

1S/C21H16/c1-13-6-7-15-12-20-17-5-3-2-4-14(17)8-9-18(20)19-11-10-16(13)21(15)19/h2-9,12H,10-11H2,1H3

InChI key

PPQNQXQZIWHJRB-UHFFFAOYSA-N

General description

3-Methylcholanthrene is a potent carcinogenic polycyclic aromatic hydrocarbon. It plays a role in the pathogenesis of human colon cancer.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Applied Microbiology, Volume 30, 258-258 (1984)
A method for measurement of nanogram quantities of 3-methylcholanthrene in stool samples.
Duane W C, et al.
Journal of Lipid Research, 25(5), 523-526 (1984)
Elin Swedenborg et al.
Molecular and cellular endocrinology, 362(1-2), 39-47 (2012-05-29)
The two estrogen receptor isoforms ERα and ERβ mediate biological effects of estrogens, but are also targets for endocrine disruptive chemicals (EDCs), compounds that interfere with hormonal signaling. 3-Methylcholanthrene (3-MC) and dioxin (TCDD) are EDCs and prototypical aryl hydrocarbon receptor
Tarquin Dorrington et al.
Aquatic toxicology (Amsterdam, Netherlands), 124-125, 106-113 (2012-09-04)
In fish there are four cytochrome P450 (CYP1) subfamilies: CYP1A, CYP1B, CYP1C, and CYP1D. Here we cloned Poecilia vivipara CYP1A, with an inferred amino acid sequence 91% identical to CYP1A from the killifish Fundulus heteroclitus, another member of the Cypriniformes
Feiyan Liu et al.
The Journal of biological chemistry, 287(30), 25530-25540 (2012-06-07)
Fas is a member of the death receptor family. Stimulation of Fas leads to induction of apoptotic signals, such as caspase 8 activation, as well as "non-apoptotic" cellular responses, notably NF-κB activation. Convincing experimental data have identified NF-κB as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service