Skip to Content
Merck
All Photos(2)

Documents

46023

Supelco

Fenthion-sulfone

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15O5PS2
CAS Number:
Molecular Weight:
310.33
Beilstein:
8144183
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1ccc(c(C)c1)S(C)(=O)=O

InChI

1S/C10H15O5PS2/c1-8-7-9(15-16(17,13-2)14-3)5-6-10(8)18(4,11)12/h5-7H,1-4H3

InChI key

ZDHYERRNXRANLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fenthion-sulfone is one of the metabolites of fenthion which is classified under the organophosphorus group of pesticides.

Application

Fenthion-sulfone may be used as an analytical reference standard for the determination of fenthion-sulfone in:
  • Fruits and vegetables and other food matrices by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and low-pressure gas chromatography-time-of-flight mass spectrometry (LP-GC-TOFMS).
  • Water samples by solid-phase extraction (SPE), liquid chromatography-triple quadrupole tandem mass spectrometry (LC-QqQ-MS/MS) and LC coupled to hybrid QTOF-MS.
  • Tea samples by QuEChERS extraction, liquid-liquid extraction (LLE) cleanup and analysis using GC-MS/MS equipped with multiple reaction monitoring (MRM) detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Streamlining sample preparation and gas chromatography-tandem mass spectrometry analysis of multiple pesticide residues in tea.
Cajka T, et al.
Analytica Chimica Acta, 743(43), 51-60 (2012)
Screening method for organophosphorus insecticides and their metabolites in olive oil samples based on headspace solid-phase microextraction coupled with gas chromatography.
Tsoutsi C, et al.
Analytica Chimica Acta, 573, 216-222 (2006)
Variability of matrix effects in liquid and gas chromatography-mass spectrometry analysis of pesticide residues after QuEChERS sample preparation of different food crops.
Kwon H, et al.
Journal of Chromatography A, 1270(43), 235-245 (2012)
High throughput analysis of 150 pesticides in fruits and vegetables using QuEChERS and low-pressure gas chromatography-time-of-flight mass spectrometry.
Koesukwiwat U, et al.
Journal of Chromatography A, 1217(43), 6692-6703 (2010)
Combined use of liquid chromatography triple quadrupole mass spectrometry and liquid chromatography quadrupole time-of-flight mass spectrometry in systematic screening of pesticides and other contaminants in water samples.
Masia A, et al.
Analytica Chimica Acta, 761(43), 117-127 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service