Skip to Content
Merck
All Photos(2)

Key Documents

45544

Supelco

Jodfenphos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8Cl2IO3PS
CAS Number:
Molecular Weight:
413.00
Beilstein:
1885795
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1cc(Cl)c(I)cc1Cl

InChI

1S/C8H8Cl2IO3PS/c1-12-15(16,13-2)14-8-4-5(9)7(11)3-6(8)10/h3-4H,1-2H3

InChI key

LFVLUOAHQIVABZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Determination of maximum permissible concentration of iodofenphos in the water reservoirs].
D P Zabulite et al.
Gigiena i sanitariia, (11)(11), 75-76 (1980-11-01)
A comparative evaluation of alfacron with commonly used insecticides for control of few crawling insects.
K K Gupta et al.
Indian journal of public health, 35(4), 119-120 (1991-10-01)
Allergic contact dermatitis from the fungicide Rondo-M and the insecticide Alfacron.
D Iliev et al.
Contact dermatitis, 36(1), 51-51 (1997-01-01)
[Control of flies and cockroaches in the zoo with Alfacron].
A Fagasiński et al.
Wiadomosci parazytologiczne, 32(4-6), 581-583 (1986-01-01)
J A Hafez
Journal of the Egyptian Society of Parasitology, 22(3), 839-849 (1992-12-01)
Five new selective formulations were tested for effectiveness, applicability and costs in controlling the housefly Musca domestica. The tested formulations are Alfacron SB, Alfacron 10 WP, Snip IRB (a.i. Azamethiphos, thiophosphorester), Baygon SB and Golden Marlin Results of laboratory studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service