Skip to Content
Merck
All Photos(1)

Key Documents

45346

Supelco

Epoxy embedding medium, hardener DDSA

for microscopy

Synonym(s):

Dodecenylsuccinic anhydride, DDSA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26O3
CAS Number:
Molecular Weight:
266.38
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
MA.02
Pricing and availability is not currently available.

grade

for microscopy

Quality Level

density

1.00 g/mL at 20 °C

application(s)

agriculture
environmental

SMILES string

CCCCCCCCC\C=C\CC1CC(=O)OC1=O

InChI

1S/C16H26O3/c1-10(2)6-11(3)7-12(4)8-13(5)14-9-15(17)19-16(14)18/h8,10-12,14H,6-7,9H2,1-5H3/b13-8+

InChI key

WBRBLKRDTHYRRV-MDWZMJQESA-N

Looking for similar products? Visit Product Comparison Guide

General description

additional information see 45359 Epoxy Embedding Medium kit

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danijela Petrovic et al.
Experimental and molecular pathology, 104(3), 199-211 (2018-05-05)
In our recent studies we have designed fullerenol/doxorubicin nanocomposite (FNP/DOX) as the new drug nanocarrier. This research has demonstrated that this novel nanocomposite has had better implications on the liver tissue in vivo (Wistar rats treated intraperitoneally), than treatment based
Ziyi Yin et al.
Autophagy, 16(5), 900-916 (2019-07-18)
Cells are faced with various stresses during their growth and development, and autophagy is a degradative process in which cells can break down their own components to recycle macromolecules and provide energy under these stresses. For pathogenic fungi that utilize

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service