Skip to Content
Merck
All Photos(1)

Key Documents

45324

Supelco

Amitrol

PESTANAL®, analytical standard

Synonym(s):

3-Amino-1,2,4-triazole, 1,2,4-Triazol-3-amine, 3-AT, Amitrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4N4
CAS Number:
Molecular Weight:
84.08
Beilstein:
107687
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

150-153 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Nc1nc[nH]n1

InChI

1S/C2H4N4/c3-2-4-1-5-6-2/h1H,(H3,3,4,5,6)

InChI key

KLSJWNVTNUYHDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianyang Chen et al.
Plant & cell physiology, 52(5), 909-921 (2011-04-08)
Inhibition of lycopene cyclization decreased the salt tolerance of the euhalophyte Salicornia europaea L. We isolated a β-lycopene cyclase gene SeLCY from S. europaea and transformed it into Arabidopsis with stable expression. Transgenic Arabidopsis on post-germination exhibited enhanced tolerance to
J H Caufield et al.
Methods (San Diego, Calif.), 58(4), 317-324 (2012-12-13)
Two-hybrid (Y2H) assays are available in a variety of different versions, including bacterial, yeast, and mammalian systems. However, even when done exclusively in yeast, multiple different host strains, vectors, reporter genes, or protocols can be used. Here we systematically compare
Ali A Ensafi et al.
Colloids and surfaces. B, Biointerfaces, 109, 45-51 (2013-04-23)
The interaction of amitrole and salmon sperm ds-DNA was studied using UV-vis and differential pulse voltammetry (DPV) at both bare and DNA-modified electrodes. Amitrole showed an oxidation peak at 0.445 V at a bare pencil graphite electrode (PGE). When ds-DNA
G Ermondi et al.
Journal of chromatography. A, 1252, 84-89 (2012-07-17)
Molecular Interaction Fields (MIFs) based descriptors can be conveniently used to characterize and compare chromatographic scales. In this study, Quantitative Structure-Retention Relationships (QSRR) for eight different chromatographic systems were obtained with VolSurf+ descriptors and Partial Least Squares (PLS). A new
Kathryn M Gauthier et al.
American journal of physiology. Renal physiology, 301(4), F765-F772 (2011-07-15)
Cytochrome P-450 metabolites of arachidonic acid, the epoxyeicosatrienoic acids (EETs) and hydrogen peroxide (H(2)O(2)), are important signaling molecules in the kidney. In renal arteries, EETs cause vasodilation whereas H(2)O(2) causes vasoconstriction. To determine the physiological contribution of H(2)O(2), catalase is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service