Skip to Content
Merck
All Photos(2)

Key Documents

44818

Supelco

Eicosane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)18CH3
CAS Number:
Molecular Weight:
282.55
Beilstein:
1700722
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

9.8 (vs air)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

220 °C/30 mmHg (lit.)

mp

35-37 °C (lit.)
37-40 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCC

InChI

1S/C20H42/c1-3-5-7-9-11-13-15-17-19-20-18-16-14-12-10-8-6-4-2/h3-20H2,1-2H3

InChI key

CBFCDTFDPHXCNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Syakti et al.
Environmental research, 96(2), 228-234 (2004-08-25)
The biotransformation activities of two hydrocarbonoclastic marine bacteria, Corynebacterium sp. and Sphingomonas sp. 2MPII, on n-eicosane and phenanthrene were investigated. During a 56-day experiment, in pure and mixed cultures, Corynebacterium sp. and Sphingomonas sp. 2MPII removed about 70% of the
B Wenclawiak et al.
Journal of biochemical and biophysical methods, 43(1-3), 197-207 (2000-06-28)
All six insecticide active ingredients in pyrethrum extract were quantified by supercritical fluid chromatography and carbon calibration. Allethrin is a suitable reference compound for carbon calibration and pyrethrins calibrations. Carbon quantification in SFC is also applied to pyrethroids (phenothrin, permethrin
Fernando J Guerrieri et al.
Proceedings. Biological sciences, 276(1666), 2461-2468 (2009-04-15)
Discriminating among individuals and rejecting non-group members is essential for the evolution and stability of animal societies. Ants are good models for studying recognition mechanisms, because they are typically very efficient in discriminating 'friends' (nest-mates) from 'foes' (non-nest-mates). Recognition in
L C van Gorkom et al.
Biochemistry, 31(3), 671-677 (1992-01-28)
The rate of formation of high-curvature intermediates or disordered cubic phases in N-methyldioleoylphosphatidylethanolamine (N-methyl-DOPE) dispersions with or without additives was studied by 31P NMR spectroscopy. In N-methyl-DOPE dispersions, both the L alpha liquid-crystalline phase and the hexagonal HII phase convert
Songpon Phadungphatthanakoon et al.
ACS applied materials & interfaces, 3(9), 3691-3696 (2011-09-03)
Existing encapsulated organic phase change materials (PCM) usually contain a shell material that possesses a poor heat storage capacity and so results in a lowered latent heat storage density of the encapsulated PCM compared to unencapsulated PCM. Here, we demonstrate

Protocols

Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service