Skip to Content
Merck
All Photos(2)

Key Documents

43999

Supelco

5-Hydroxyomeprazole

analytical standard

Synonym(s):

4-Methoxy-6-{[(5-methoxy-1H-benzimidazol-2-yl)sulfinyl]methyl}-5-methyl-3-pyridinemethanol, 5-Methoxy-2-{[(5-hydroxymethyl-4-methoxy-3-methyl-2-pyridinyl)methyl]sulfinyl}-1H-benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3O4S
CAS Number:
Molecular Weight:
361.42
Beilstein:
8439890
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=S(CC1=NC=C(CO)C(OC)=C1C)C2=NC3=CC(OC)=CC=C3N2

InChI

1S/C17H19N3O4S/c1-10-15(18-7-11(8-21)16(10)24-3)9-25(22)17-19-13-5-4-12(23-2)6-14(13)20-17/h4-7,21H,8-9H2,1-3H3,(H,19,20)

InChI key

CMZHQFXXAAIBKE-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valerie A Frerichs et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 824(1-2), 71-80 (2005-07-20)
A method has been developed and validated for the quantitation of midazolam, alphahydroxy-midazolam, omeprazole, and hydroxyomeprazole from one 250 microL sample of human plasma using high performance liquid chromatography coupled to tandem mass spectrometry. The method was validated for a
J Olsson et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 875(1), 329-332 (2008-06-25)
The present paper demonstrates the enantiomeric separation of omeprazole and its metabolite 5-hydroxyomeprazole performed with open tubular capillary electrochromatography (OT-CEC). The protein avidin was used as the chiral selector. Avidin was immobilized by a Schiffs base type of reaction where
Marcelo Ribani et al.
Journal of chromatography. A, 1156(1-2), 201-205 (2007-02-03)
To guarantee that an analytical procedure gives reliable, exact and interpretable information about a sample, it must be validated. Two ambiguous parameters are detection limit and quantification limit. The determination of these limits is still of great concern and there
B L Chen et al.
Journal of clinical pharmacology, 49(5), 574-581 (2009-04-29)
This study explores the impact of clopidogrel on the pharmacokinetics of omeprazole related to CYP2C19 genetic polymorphisms. Twelve healthy volunteers (6 CYP2C19*1/*1, 5 CYP2C19*2/*2, and 1 CYP2C19*2/*3) are enrolled in a 2-phase randomized crossover trial. In each phase, the volunteers
Y S Pang et al.
British journal of clinical pharmacology, 58(3), 332-335 (2004-08-26)
Impaired S-mephenytoin 4'-hydroxylation is a well-described genetic polymorphism affecting drug metabolism in humans. Although ethnic differences in its distribution of polymorphism has been described, it is not known whether there is an ethnic heterogeneity of the structure and expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service