42235
BDE No 99
analytical standard
Synonym(s):
1,2,4-Tribromo-5-(2,4-dibromophenoxy)benzene, 2,2′,4,4′,5-PentaBDE, 2,2′,4,4′,5-Pentabromodiphenyl ether, BDE 99, PBDE 99
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
application(s)
environmental
format
neat
SMILES string
Brc1c(cc(c(c1)Br)Oc2c(cc(cc2)Br)Br)Br
InChI
1S/C12H5Br5O/c13-6-1-2-11(9(16)3-6)18-12-5-8(15)7(14)4-10(12)17/h1-5H
InChI key
WHPVYXDFIXRKLN-UHFFFAOYSA-N
Recommended products
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - STOT RE 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service