Skip to Content
Merck
All Photos(2)

Documents

39319

Supelco

2,5-Dihydroxybenzoic acid

matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure

Synonym(s):

2,5-DHBA, DHB, Gentisic acid, Hydroquinonecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
2209119
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.5% (HPLC)

analyte functional class(es)

polymers

analyte chemical class(es)

glycans, lipids, organic molecules, peptides, proteins

technique(s)

MALDI-MS: suitable

mp

203-207 °C
204-208 °C (lit.)

solubility

methanol: 10 mg/mL, clear

cation traces

Al: ≤1 mg/kg
Ba: ≤1 mg/kg
Ca: ≤1 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤1 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

SMILES string

OC(=O)c1cc(O)ccc1O

InChI

1S/C7H6O4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H,(H,10,11)

InChI key

WXTMDXOMEHJXQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Quantitative MALDI-MS and Imaging of Fungicide Pyrimethanil in Strawberries with 2-Nitrophloroglucinol as an Effective Matrix.: This research highlights the application of 2,5-Dihydroxybenzoic acid (DHB) as an effective matrix for MALDI-MS imaging, providing enhanced detection sensitivity for fungicides in agricultural samples. The study showcases DHB′s role in improving the quantification and spatial resolution of pesticide residues within fruit matrices, demonstrating its utility in food safety and chemical analysis (Liang et al., 2024).

  • A maize enzyme from the 2-oxoglutarate-dependent oxygenase family with unique kinetic properties, mediates resistance against pathogens and regulates senescence.: Utilizing 2,5-Dihydroxybenzoic acid in enzyme kinetics studies, this paper reveals its critical role in modulating plant defense mechanisms against pathogens. The study provides insights into the biochemical pathways influenced by DHB, underscoring its importance in plant biochemistry and resistance breeding (Casati et al., 2024).

  • Unveiling the distribution of free and bound phenolic acids, flavonoids, anthocyanins, and proanthocyanidins in pigmented and non-pigmented rice genotypes.: This comprehensive analysis employs 2,5-Dihydroxybenzoic acid to investigate the phenolic content in different rice varieties, highlighting its role in enhancing the detection of health-beneficial compounds. The research supports DHB′s application in nutritional studies and crop quality assessments (Thulasinathan et al., 2024).

  • Novel Small Molecule Matrix Screening for Simultaneous MALDI Mass Spectrometry Imaging of Multiple Lipids and Phytohormones.: Leveraging the chemical properties of 2,5-Dihydroxybenzoic acid, this study introduces a novel matrix for MALDI-MS imaging, enhancing the simultaneous visualization of lipids and phytohormones in biological tissues. It highlights the versatility of DHB in facilitating complex biochemical analyses (Shi et al., 2024).

  • 2-Hydroxy-5-nitro-3-(trifluoromethyl)pyridine as a Novel Matrix for Enhanced MALDI Imaging of Tissue Metabolites.: This research demonstrates the use of DHB-based matrices to improve the MALDI imaging of metabolites, offering greater sensitivity and specificity in clinical diagnostics and metabolic studies. The findings contribute to the broader application of DHB in analytical chemistry (Wang et al., 2024).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V. K. Ahluwalia
Intermediates for Organic Synthesis, 78-79 (2005)
Ya-Shiuan Lin et al.
Analytical chemistry, 74(22), 5793-5798 (2002-12-05)
This work presents a novel method for direct desorption/ ionization of analytes from sol-gel-derived film. 2,5-Dihydroxy benzoic acid (DHB), a common MALDI matrix, was incorporated into a sol-gel polymeric structure. The sol-gel-derived DHB thin film can assist the mass analysis
F Bochner et al.
Clinical pharmacology and therapeutics, 30(2), 266-275 (1981-08-01)
Single oral doses of aspirin (ASA, 1,500 mg), sodium salicylate (NaSA, 1,500 mg, 1,200 mg), and salicyluric acid (SUA, 500 mg) were given to five subjects. Serial plasma and urine samples were collected for 24 hr (plasma) and up to
Mandana Hunter et al.
Journal of neurochemistry, 146(5), 613-630 (2018-05-29)
The human subventricular zone (SVZ) has a defined cytological and neurochemical architecture, with four constituent laminae that act in concert to support its neurogenic activity. Lipidomic specialisation has previously been demonstrated in the neurologically normal human SVZ, with enrichment of
J N Buskin et al.
Clinical chemistry, 28(5), 1200-1203 (1982-05-01)
Detailed performance specifications are given for a specific and sensitive liquid-chromatographic assay for aspirin, salicylic acid, and salicyluric acid in plasma. The sensitivity of this method for aspirin (50 micrograms/L) is 10-fold that of previous methods, so that concentrations of

Articles

One of the most important aspects of our ultra-pure MALDI matrix substances is their ability to dissolve rapidly and completely; a brief vortex mixing is typically sufficient.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service