Skip to Content
Merck
All Photos(2)

Documents

38800

Supelco

Malachite Green chloride

analytical standard

Synonym(s):

N,N,N′,N′-Tetramethyl-4,4′-diaminotriphenylcarbenium chloride, Basic Green 4, Malachite Green

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25ClN2
CAS Number:
Molecular Weight:
364.91
Colour Index Number:
42000
Beilstein:
3580148
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Cl-].CN(C)c1ccc(cc1)C(\c2ccccc2)=C3\C=CC(\C=C3)=[N+](\C)C

InChI

1S/C23H25N2.ClH/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;/h5-17H,1-4H3;1H/q+1;/p-1

InChI key

FDZZZRQASAIRJF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Malachite Green chloride (MG), a neat reference standard, is provided for the analyte detection, food color quality, and safety control of your products! Malachite Green is a low-cost, highly efficient, cationic triphenylmethane dye with high efficacy for treating fungal, bacterial, and parasitic infections. MG is also employed in paper, textile, acrylic industries, and as an anthelmintic agent, food and color additive, medical disinfectant, etc.
Malachite green chloride is a triphenylmethane dye, which generally can find applications in aquaculture and highly effective against fungal infections.

Application

Malachite Green chloride may be used as an analytical standard to investigate its carcinogenic nature in the animal matrix. The techniques, proton nuclear magnetic resonance (1H), carbon-13 nuclear magnetic resonance (13C NMR), high-performance liquid chromatography (HPLC), and gas chromatography (GC) are used to confirm the identity of the material and to determine the purity of malachite green chloride.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of malachite green (MG) and its major metabolite, leucomalachite green (LMG), in two human cell lines
Stammati A, et al.
Toxicology in vitro, 19(7), 853-858 (2005)
Carcinogenicity of malachite green chloride and leucomalachite green in B6C3F1 mice and F344 rats
Culp JS, et al.
Food And Chemical Toxicology, 44(8), 1204-1212 (2006)
Crystall M D Swarbrick et al.
Nucleic acids research, 45(22), 12904-12920 (2017-11-23)
The protein-RNA interactions within the flavivirus replication complex (RC) are not fully understood. Our structure of dengue virus NS3 adenosine triphosphatase (ATPase)/helicase bound to the conserved 5' genomic RNA 5'-AGUUGUUAGUCU-3' reveals that D290 and R538 make specific interactions with G2
Yoshinobu Uemoto et al.
BMC genomics, 18(1), 874-874 (2017-11-15)
Umami is a Japanese term for the fifth basic taste and is an important sensory property of beef palatability. Inosine 5'-monophosphate (IMP) contributes to umami taste in beef. Thus, the overall change in concentration of IMP and its degradation products
Effects of malachite green (MG) and its major metabolite, leucomalachite green (LMG), in two human cell lines
Stammati A,et al.
Toxicology in vitro, 19, 853-858 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service