Skip to Content
Merck
All Photos(1)

Documents

36663

Supelco

2,4′-DDE solution

100 μg/mL in methanol, PESTANAL®, analytical standard

Synonym(s):

2-(2-Chlorophenyl)-2-(4-chlorophenyl)-1,1-dichloroethene solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8Cl4
CAS Number:
Molecular Weight:
318.03
Beilstein:
1977640
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)\C(=C(\Cl)Cl)c2ccccc2Cl

InChI

1S/C14H8Cl4/c15-10-7-5-9(6-8-10)13(14(17)18)11-3-1-2-4-12(11)16/h1-8H

InChI key

ZDYJWDIWLRZXDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sergio A Villalobos et al.
Chemosphere, 53(8), 819-826 (2003-09-25)
The toxicity of o,p'-DDE (1,1-dichloro-2-(p-chlorophenyl)-2-(o-chlorophenyl) ethylene) was evaluated in embryos of medaka (Oryzias latipes) following a one time exposure via nanoinjection. Medaka eggs (early gastrula) were injected with 0.5 nl of triolein (vehicle control) or 0.5 nl of 4 graded
A K Wójtowicz et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 58(4), 873-885 (2008-01-16)
We evaluated impact of DDT isomers, o, p'- DDT [1, 1-dichloro-2, 2-bis (p, p'-chlorophenyl) ethylene] and p, p'-DDT [1, 1, 1-trichloro-2, 2-bis (p-chlorophenyl) ethane], and their metabolites, o, p'-DDE and p, p'-DDE, on ovarian steroidogenesis. All these compounds, except for
Yanjing Wu et al.
Reproduction (Cambridge, England), 131(2), 299-310 (2006-02-03)
Following our previous demonstration that p,p'-DDE (dichlorodiphenylchloroethylene), at environmentally relevant concentrations, can rapidly increase intracellular calcium [Ca2+]i concentrations in human granulosa-lutein cells, we examined whether other pesticides, such as Kepone, o,p-DDE and methoxychlor, have similar effects. Cultured human granulosa-lutein cells
Anna K Wójtowicz et al.
Polish journal of pharmacology, 56(4), 465-472 (2004-11-03)
The aim of the presented study was to compare the effect of o,p'-DDT [1,1-dichloro-2,2-bis-(p,p'-chlorophenyl)-ethylene] and p,p'-DDT [1,1,1-trichloro-2,2-bis-(p-chlorophenyl)-ethane] and their metabolites DDE and DDD on estradiol secretion by ovarian follicles, the target organs of environmental estrogens. Theca interna (Tc) and granulosa
Lori K Davis et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 149(4), 507-514 (2008-12-23)
Effects of two endocrine disruptors, o,p'-DDE and heptachlor, and 17beta-estradiol (E(2)) on vitellogenin (Vg) and the growth hormone (GH)/insulin-like growth factor-I (IGF-I) axis were examined in male tilapia. In the first experiment, fish were given 5 weekly injections of either

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service