Skip to Content
Merck
All Photos(1)

Documents

36524

Supelco

8-Quinolinol

PESTANAL®, analytical standard

Synonym(s):

8-Hydroxyquinoline, 8-Oxychinolin, Oxine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
Beilstein:
114512
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

98-100% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

72.5-74.0 °C

antibiotic activity spectrum

fungi

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

Oc1cccc2cccnc12

InChI

1S/C9H7NO/c11-8-5-1-3-7-4-2-6-10-9(7)8/h1-6,11H

InChI key

MCJGNVYPOGVAJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Isabel Fernández-Bachiller et al.
Journal of medicinal chemistry, 53(13), 4927-4937 (2010-06-16)
Tacrine and PBT2 (an 8-hydroxyquinoline derivative) are well-known drugs that inhibit cholinesterases and decrease beta-amyloid (Abeta) levels by complexation of redox-active metals, respectively. In this work, novel tacrine-8-hydroxyquinoline hybrids have been designed, synthesized, and evaluated as potential multifunctional drugs for
Bhimashankar Mitkari et al.
Experimental neurology, 239, 158-162 (2012-10-13)
Cell therapies from various sources have been under intense research in stroke. Efficient homing of the cells to the injured brain without complications is necessary to realize the therapeutic potential of cell therapy. Intra-arterial (IA) infusion of cells bypasses the
Steve Comby et al.
Inorganic chemistry, 51(19), 10158-10168 (2012-09-15)
A novel near-infrared (NIR) emissive lanthanide-based zinc sensor was designed, based on the self-assembly in aqueous solution between the nonemissive coordinatively unsaturated Yb(III) cyclen complex 2·Yb and the sulfonated 8-hydroxyquinoline (8-HQS) chromophore, which was employed as a sensitizing antenna. The
Mahmud Tareq Hassan Khan et al.
Journal of medicinal chemistry, 52(1), 48-61 (2008-12-17)
In the present work, 22 compounds of the U.S. NCI compound library (size 273K) were identified as putative thermolysin binders by structure based virtual screening with the ICM software (ICM-VLS). In vitro competitive binding assays confirmed that 12 were thermolysin
G L Apostoli et al.
Journal of thrombosis and haemostasis : JTH, 12(11), 1880-1889 (2014-08-29)
Nitric oxide (NO) is a critical negative regulator of platelets that is implicated in the pathology of thrombotic diseases. Platelets generate NO, but the presence and functional significance of NO synthase (NOS) in platelets is unclear. Inorganic nitrate/nitrite is increasingly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service