Skip to Content
Merck
All Photos(2)

Key Documents

35833

Supelco

4-Chloro-2-methylphenol

PESTANAL®, analytical standard

Synonym(s):

4-Chloro-o-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)OH
CAS Number:
Molecular Weight:
142.58
Beilstein:
1906684
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

220-225 °C (lit.)

mp

43-46 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cc(Cl)ccc1O

InChI

1S/C7H7ClO/c1-5-4-6(8)2-3-7(5)9/h2-4,9H,1H3

InChI key

RHPUJHQBPORFGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janne Juntunen et al.
Analytica chimica acta, 1127, 269-281 (2020-08-18)
Current theoretical, two compartment description of integrative passive sampling is renewed to establish a three-compartment model. The developed theoretical description includes external chemical conditions near the receiving phase, conditions inside the receiving phase and the chemically bonded compartments. New variable
E Lynge
British journal of cancer, 52(2), 259-270 (1985-08-01)
The purpose of this cohort study is to shed further light on the potential carcinogenic effect indicated by a Swedish case control study of the 2,4-dichlorophenol and 4-chloro-ortho-cresol based phenoxy herbicides, unlikely to be contaminated with 2,3,7,8-tetrachlorodibenzo-p-dioxin (2,3,7,8-TCDD). In the
Identification of 4-chloro-2-methylphenol as a soil degradation product of ring-labelled [14C]mecoprop.
A E Smith
Bulletin of environmental contamination and toxicology, 34(5), 656-660 (1985-05-01)
K H Oh et al.
FEMS microbiology letters, 63(2-3), 141-146 (1991-04-15)
The concurrent bacterial degradation of 2-(2-methyl-4-chlorophenoxy)propionic acid and 2,4-dichlorophenoxyacetic acid was studied using a stirred tank reactor and a bacterial culture which had been originally derived by enrichment with MCPP. High pressure liquid chromatographic methodology was used to measure both
U Lechner et al.
Biodegradation, 6(2), 83-92 (1995-06-01)
The Gram-negative strain S1, isolated from activated sludge, metabolized 4-chloro-2-methylphenol by an inducible pathway via a modified ortho-cleavage route as indicated by a transiently secreted intermediate, identified as 2-methyl-4-carboxymethylenebut-2-en-4-olide by gas chromatography/mass spectrometry. Beside 4-chloro-2-methylphenol only 2,4-dichlorophenol and 4-chlorophenol were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service