Skip to Content
Merck
All Photos(1)

Key Documents

34322

Supelco

Chlorosulfuron

PESTANAL®, analytical standard

Synonym(s):

1-(2-Chlorophenylsulfonyl)-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-urea, Chlorsulfuron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12ClN5O4S
CAS Number:
Molecular Weight:
357.77
Beilstein:
577255
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2Cl)n1

InChI

1S/C12H12ClN5O4S/c1-7-14-10(17-12(15-7)22-2)16-11(19)18-23(20,21)9-6-4-3-5-8(9)13/h3-6H,1-2H3,(H2,14,15,16,17,18,19)

InChI key

VJYIFXVZLXQVHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorsulfuron belongs to the class of sulfonylurea herbicides.

Application

Chlorosulfuron may be used as an analytical reference standard for the determination of chlorosulfuron in:
  • Environmental water samples by magnetic solid phase extraction (MSPE) using multi-walled carbon nanotubes adsorbents combined with high performance liquid chromatography-diode-array-detection (HPLC-DAD).
  • Soya milk samples by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and HPLC-DAD as well as HPLC combined with triple quadrupole tandem mass spectrometry (QqQ-MS/MS).
  • Honeybees by dispersive SPE followed by liquid and gas chromatography combined with tandem mass spectrometry (LC-MS/MS and GC-MS/MS) with electrospray ionization (ESI), and multiple reaction monitoring (MRM) detection.
  • Fruits and vegetable samples by methanolic extraction and ultra-HPLC in conjunction with ESI-MS/MS and MRM detection.
  • Cereals by QuEChERS extraction and HPLC coupled to ESI-MS/MS with selected reaction monitoring (SRM) detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of four sulfonylurea herbicides in cereals using modified Quick, Easy, Cheap, Effective, Rugged, and Safe sample preparation method coupled with liquid chromatography-tandem mass spectrometry.
Ni Y, et al.
Journal of Chromatography A, 1537, 27-34 (2018)
Simple, cost-effective and sensitive liquid chromatography diode array detector method for simultaneous determination of eight sulfonylurea herbicides in soya milk samples.
Rejczak T and Tuzimski T
Journal of Chromatography A, 1473, 56-65 (2016)
Qing-fu Ye et al.
Journal of environmental sciences (China), 16(2), 262-267 (2004-05-13)
The bioavailability of bound residue (BR) derived from 14C-labeled chlorsulfuron in soil and effect of the main components of the BR on growth of rape (brassica napus) and rice (Oryza sativa L.) were investigated. The results showed that the BR
René Ruiter et al.
Plant molecular biology, 53(5), 675-689 (2004-03-11)
In this study we tested the performance of chimeraplasts, chimeric RNA/DNA oligonucleotides, for the creation of directed changes in chromosomal sequences in tobacco and oilseed rape. As target genes for chimeraplasty, the endogenous als gene and two transgenes, bar and
S Slavov et al.
Transgenic research, 14(3), 273-278 (2005-09-09)
Broomrape (Orobanche ramosa L.) is the most important parasitic plant that infests tobacco (Nicotiana tabacum L.). Chemical treatment of the soil is not effective and crop rotation is not acceptable to solve this problem because of the long viability period

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service