Skip to Content
Merck
All Photos(1)

Key Documents

34037

Supelco

Ochratoxin A solution

10 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18ClNO6
CAS Number:
Molecular Weight:
403.81
Beilstein:
8169012
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

methanol: soluble 5 mg/mL, clear, yellow(lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

C[C@@H]1Cc2c(Cl)cc(c(O)c2C(=O)O1)C(=O)N[C@@H](Cc3ccccc3)C(O)=O

InChI

1S/C20H18ClNO6/c1-10-7-12-14(21)9-13(17(23)16(12)20(27)28-10)18(24)22-15(19(25)26)8-11-5-3-2-4-6-11/h2-6,9-10,15,23H,7-8H2,1H3,(H,22,24)(H,25,26)/t10-,15+/m1/s1

InChI key

RWQKHEORZBHNRI-BMIGLBTASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial
Ochratoxin is a mycotoxin, produced by genus Aspergillus and Penicillium. Ochratoxin producers are widely distributed in foods such as cereals.[1] Ochratoxins are isocoumarin derivatives.[2] A quantitative detection method for ochratoxin A by using aptamer (single-stranded oligonucleotides selected in vitro to bind to molecular targets) is reported.[3]

Application

Ochratoxin A (OTA) was used in quantification of OTA in table wine by immunoaffinity column clean-up and high-performance liquid chromatography.[1] It was used as analytical standard in sample clean-up method based on immuno-ultrafiltration for the analysis of OTA in cereals.[2] It was used as analytical standard to study the influence of OTA on the rat embryonic neural cells cultured in high density “micromass cultures”.[4]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iwona Wilk-Zasadna et al.
International journal of molecular sciences, 10(1), 37-49 (2009-04-01)
Embryonic midbrain micromass cultures were exposed for five days to ochratoxin A (OTA) at seven concentrations (ranging from 0.16 to 10 microg/mL). Cell viability was assessed in neutral red uptake test (NRU), and differentiation - by immunoenzymatic determination of structural
Elisabeth Viktoria Reiter et al.
Analytical and bioanalytical chemistry, 400(8), 2615-2622 (2011-04-05)
The paper presents a new sample clean-up method based on immuno-ultrafiltration for the analysis of ochratoxin A in cereals. In contrast to immunoaffinity chromatography, in immuno-ultrafiltration, the antibodies are used in non-immobilised form. Ochratoxin A was extracted with ACN/water (60/40
Jorge A Cruz-Aguado et al.
Journal of agricultural and food chemistry, 56(22), 10456-10461 (2008-11-06)
This work describes the identification of an aptamer that binds with high affinity and specificity to ochratoxin A (OTA), a mycotoxin that occurs in wheat and other foodstuffs, and a quantitative detection method for OTA based on the use of
A Visconti et al.
Journal of chromatography. A, 864(1), 89-101 (2000-01-12)
A new and accurate method to quantify ochratoxin A (OA) in table wine has been developed. The method uses commercial immunoaffinity columns for clean-up and high-performance liquid chromatography (HPLC) with fluorescence detection for quantification of the toxin. Wine was diluted
Donata Lerda et al.
Journal of AOAC International, 96(2), 331-340 (2013-06-19)
A collaborative study was conducted to validate an analytical method for the determination of ochratoxin A (OTA) in licorice (root powder) and licorice extracts (paste and powder). Contents of OTA ranged from 26 to 141 microg/kg and from 8 to

Articles

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

Related Content

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Please I would like to know if the information contained in the certificate "The minimum recommended amount of subsample for all application types is 1 mL" is correct. Normally, it is allowed to withdraw vol. of up to 10 microliters from the standard

    1 answer
    1. The minimum recommended sub-sample is 1mL. The ampoule should be used once and not stored and re-sampled. Accuracy is optimized by transferring the maximum amount of standard solution.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service