Skip to Content
Merck
All Photos(1)

Documents

33999

Supelco

Rifaximin

VETRANAL®, analytical standard

Synonym(s):

4-Deoxy-4′-methylpyrido[1′,2′-1,2]imidazo[5,4-c]rifamycin SV, Rifacol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H51N3O11
CAS Number:
Molecular Weight:
785.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)C(C)=C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@H](O)[C@H](C)[C@H]([C@H]1C)C(=O)OC)c5c(nc6cc(C)ccn56)c4c3C2=O

InChI

1S/C43H51N3O11/c1-19-14-16-46-27(18-19)44-32-29-30-37(49)25(7)39-31(29)40(51)43(8,57-39)56-17-15-26(54-9)22(4)28(42(53)55-10)23(5)36(48)24(6)35(47)20(2)12-11-13-21(3)41(52)45-33(34(32)46)38(30)50/h11-18,20,22-24,26,28,35-36,47-50H,1-10H3,(H,45,52)/b12-11+,17-15+,21-13-/t20-,22+,23-,24-,26+,28+,35-,36-,43+/m1/s1

InChI key

HIYLTQREEOINNF-HTEWPBCCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rifaximin may have a dramatic effect on hepatic encephalopathy.
Christopher Lamb et al.
Clinical medicine (London, England), 12(5), 489-490 (2012-10-30)
E Mattila et al.
Alimentary pharmacology & therapeutics, 37(1), 122-128 (2012-10-26)
Clostridium difficile can cause severe antibiotic-associated colitis. Conventional treatments with metronidazole and vancomycin improve symptoms, but after discontinuation of treatment, C. difficile infection (CDI) recurs in a number of patients. Rifaximin is a rifamycin-based non-systemic antibiotic that has effect against
Vishesh Kothary et al.
Antimicrobial agents and chemotherapy, 57(2), 811-817 (2012-11-28)
Escherichia coli is implicated in the pathogenesis of inflammatory bowel disease (IBD). Rifaximin, a nonabsorbable derivative of rifampin effective against E. coli, improves symptoms in mild-to-moderate IBD. However, rifaximin resistance can develop in a single step in vitro. We examined
Rosacea and small intestinal bacterial overgrowth: prevalence and response to rifaximin.
Leonard B Weinstock et al.
Journal of the American Academy of Dermatology, 68(5), 875-876 (2013-04-23)
Guy W Neff et al.
Journal of clinical gastroenterology, 47(2), 188-192 (2013-01-15)
The purpose of this study was to assess the incidence of Clostridium difficile infection in patients who received rifaximin for the treatment of hepatic encephalopathy (HE). Medical charts of patients who received rifaximin for the treatment of HE were reviewed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service