Skip to Content
Merck
All Photos(1)

Key Documents

33482

Sigma-Aldrich

1,8-Diazabicyclo[5.4.0]undec-7-ene

puriss., ≥99.0% (GC)

Synonym(s):

2,3,4,6,7,8,9,10-Octahydropyrimidol[1,2-a]azepine, DBU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N2
Molecular Weight:
152.24
Beilstein:
508906
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

7.1 hPa ( 37.7 °C)

Quality Level

grade

puriss.

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.523

pH

(12,8 at 10 g/l at 20 °C)

bp

115 °C/11 mmHg (lit.)

mp

-70  °C

density

1.019 g/mL at 20 °C (lit.)

SMILES string

C1CCN2CCCN=C2CC1

InChI

1S/C9H16N2/c1-2-5-9-10-6-4-8-11(9)7-3-1/h1-8H2

InChI key

GQHTUMJGOHRCHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,8-Diazabicyclo[5.4.0]undec-7-ene can be used:
  • To mediate the aza-Michael addition of nitrogen nucleophiles to α,β-unsaturated carbonyl compounds.
  • As a nucleophilic catalyst for the esterification of carboxylic acid with dimethyl carbonate.
  • As a bidentate ligand in copper (I) bromide mediated atom transfer radical polymerization of methyl methacrylate.
  • As an efficient catalyst for the transesterification of dimethyl carbonate with glycerol.
Amidine base used for dehydrohalogenation reactions to olefinscitation; Further important applications are e.g.: esterification of carboxylic acids with alkyl halidescitationcitation; Alkylation and acylation of active methylene compoundscitation

Other Notes

Amidine base used for dehydrohalogenation reactions to olefins; Further important applications are e.g.: esterification of carboxylic acids with alkyl halides; Alkylation and acylation of active methylene compounds

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F

Flash Point(C)

116 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N. Ono et al.
Bulletin of the Chemical Society of Japan, 51, 2401-2401 (1978)
C.G. Rao
Organic preparations and procedures international, 12, 225-225 (1980)
N. Ono et al.
Bulletin of the Chemical Society of Japan, 52, 1716-1716 (1979)
H. Oediger et al.
Synthesis, 591-591 (1972)
1, 8-Diazabicyclo [5.4. 0] undec-7-ene (DBU) as ligand for atom transfer radical polymerization (ATRP)
Fournier D, et al.
European Polymer Journal, 41(7), 1576-1581 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service