Skip to Content
Merck
All Photos(2)

Key Documents

32553

Supelco

Tinidazole

VETRANAL®, analytical standard

Synonym(s):

1-[2-(Ethylsulfonyl)ethyl]-2-methyl-5-nitroimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13N3O4S
CAS Number:
Molecular Weight:
247.27
Beilstein:
618182
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCS(=O)(=O)CCn1c(C)ncc1[N+]([O-])=O

InChI

1S/C8H13N3O4S/c1-3-16(14,15)5-4-10-7(2)9-6-8(10)11(12)13/h6H,3-5H2,1-2H3

InChI key

HJLSLZFTEKNLFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tinidazole belongs to the family of nitroimidazole compounds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tinidazole may be used as an analytical reference standard for the determination of the analyte in:
  • Tablet formulations and human serum samples by reversed phase-high performance liquid chromatography (RP-HPLC).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Separation and assay o f antiprotozoal imidazole derivatives (metronidazole, tinidazole and secnidazole) by RP-HPLC.
Alhalabi Z, et al.
International Journal of Pharmaceutical Sciences Review and Research, 13(1), 13-18 (2012)
Multi-residue method for the detection of veterinary drugs in distillers grains by liquid chromatography-Orbitrap high resolution mass spectrometry.
Kaklamanos G, et al.
Journal of Chromatography A, 1322, 38-48 (2013)
C E Nord et al.
Infection, 11(1), 54-60 (1983-01-01)
Tinidazole is a 5-nitroimidazole with selective activity against anaerobic bacteria and protozoa. It is bactericidal at low concentrations and its spectrum covers most anaerobic bacteria and some capnophilic microorganisms. Anaerobic bacteria known to be resistant to tinidazole include anaerobic streptococci
Hyuk Soon Choi et al.
World journal of gastroenterology, 18(19), 2377-2382 (2012-06-02)
To compare the effectiveness of sequential therapy for Helicobacter pylori (H. pylori) infection with that of triple therapy of varying durations. The 460 patients enrolled in this study had H. pylori-associated gastritis or a gastric or duodenal ulcer. After screening
Simultaneous estimation of ciprofloxacin hydrochloride, ofloxacin, tinidazole and ornidazole by reverse phase-high performance liquid chromatography.
Singh R, et al.
Eurasian Journal of Analytical Chemistry, 4(2), 161-167 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service