Recommended Products
Quality Level
Assay
96%
form
powder
composition
Carbon: 58.6-64.2%
Nitrogen: 18.6-20.4%
technique(s)
titration: suitable
color
brown-red
mp
192-202 °C (lit.)
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
Oc1ccc(\N=N\c2ccccn2)c(O)c1
InChI
1S/C11H9N3O2/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11/h1-7,15-16H/b14-13+
InChI key
RJNYNDHYSJRRDW-BUHFOSPRSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4-(2-Pyridylazo)resorcinol (PAR) is a monosodium salt(1), dibasic acid that forms the protonated complexes with most metal ions.
Application
4-(2-Pyridylazo)resorcinol (PAR) serves as a metallochromic indicator and is suitable as a chromogenic agent for the quantitative determination of over 50 elements.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Biochemistry, 44(12), 4636-4647 (2005-03-23)
The inducible isoform of nitric oxide synthase (iNOS) and three zinc tetrathiolate mutants (C104A, C109A, and C104A/C109A) were expressed in Escherichia coli and purified. The mutants were found by ICP-AES and the zinc-specific PAR colorimetric assay to be zinc free
Polymers, 13(3) (2021-02-06)
In this study, X-ray photoelectron spectroscopy (XPS) was used to study chitosan-graphene oxide (chitosan-GO) incorporated with 4-(2-pyridylazo)resorcinol (PAR) and cadmium sulfide quantum dot (CdS QD) composite thin films for the potential optical sensing of cobalt ions (Co2+). From the XPS
Marine drugs, 17(8) (2019-07-28)
In this study, three synthetic zinc-chelating peptides (ZCPs) derived from sea cucumber hydrolysates with limited or none of the common metal-chelating amino-acid residues were analyzed by flame atomic absorption spectroscopy, circular dichroism spectroscopy, size exclusion chromatography, zeta-potential, Fourier transform infrared
Dalton transactions (Cambridge, England : 2003), 39(5), 1366-1374 (2010-01-28)
Literature reports of the efficacy of para-sulfonatocalix[6]- and calix[8]-arenes as U(vi) complexants indicated that they might be useful for in vivo chelation of the novel therapeutic alpha-emitter (230)U. We have studied the complexation of U(vi) with para-sulfonatocalix[6]arene and para-sulfonatocalix[8]arene by
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 32(3), 521-532 (2019-02-14)
The biosynthesis of the dinuclear metal cluster at the active sites of the [NiFe]-hydrogenase enzymes is a multi-step process executed by a suite of accessory proteins. Nickel insertion during maturation of Escherichia coli [NiFe]-hydrogenase 3 is achieved by the metallochaperones
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service