Skip to Content
Merck
All Photos(1)

Key Documents

32091

Supelco

Sulfamonomethoxine

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(6-methoxy-4-pyrimidinyl)benzenesulfonamide, 4-Methoxy-6-sulfanilamidopyrimidine, N1-(6-Methoxy-4-pyrimidinyl)sulfanilamide, SMM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N4O3S
CAS Number:
Molecular Weight:
280.30
Beilstein:
621128
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤3.0% water

application(s)

clinical testing

format

neat

SMILES string

COc1cc(NS(=O)(=O)c2ccc(N)cc2)ncn1

InChI

1S/C11H12N4O3S/c1-18-11-6-10(13-7-14-11)15-19(16,17)9-4-2-8(12)3-5-9/h2-7H,12H2,1H3,(H,13,14,15)

InChI key

WMPXPUYPYQKQCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide
Sulfamonomethoxine is classified under the sulfonamide group of compounds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfamonomethoxine may be used as an analytical reference standard for the determination of sulfamonomethoxine in:
  • Tissues, milk and eggs of food-producing animals by ultrasound-assisted extraction (UAE) and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).
  • Food samples of animal origin by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction procedure, SOSLE (salting-out supported liquid extraction) method and nanoflow liquid chromatography high resolution mass spectrometry (LC-HRMS).
  • Bovine muscles by hydrophilic interaction liquid chromatography (HILIC) combined with LC-MS/MS.
  • Commercial eggs by liquid-liquid extraction (LLE), hybrid solid-phase extraction (SPE) and LC-MS/MS.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue method for the simultaneous determination of veterinary medicinal products, feed additives and illegal dyes in eggs using liquid chromatography-tandem mass spectrometry.
Piatkowska M, et al.
Food Chemistry, 197, 571-580 (2016)
Analysis of trace levels of sulfonamides in fish tissue using micro-scale pipette tip-matrix solid-phase dispersion and fast liquid chromatography tandem mass spectrometry.
Shen Q, et al.
Food Chemistry, 194, 508-515 (2016)
Jingchun Yan et al.
Journal of hazardous materials, 186(2-3), 1398-1404 (2011-01-18)
Iron oxide magnetic nanoparticles (Fe(3)O(4) MNPs) can effectively activate persulfate anions (S(2)O(8)(2-)) to produce sulfate free radicals (SO(4)(-)), which are a powerful oxidant with promising applications to degrade organic contaminants. The kinetics of sulfamonomethoxine (SMM) degradation was studied in the
Kunihiro Kishida et al.
Journal of chromatography. A, 1028(1), 175-177 (2004-02-19)
A hazardous-chemical free method for simultaneous determination of sulfamonomethoxine (SMM), sulfadimethoxine (SDM), and their N4-acetyl metabolites in raw milk using shielded column liquid chromatography is developed. The target analytes are extracted by mixing with ethanol-acetic acid (97:3, v/v) followed by
Hiromi Ito et al.
Carbohydrate research, 338(16), 1621-1639 (2003-07-23)
Systematic synthesis and myelin-associated glycoprotein (MAG)-binding activity of novel sulfated GM1b analogues structurally related to Chol-1 (alpha-series) gangliosides, high-affinity ligands for neural siglecs, are described. The suitably protected gangliotriose derivatives, 2-(trimethylsilyl)ethyl 2-acetamido-2-deoxy-6-O-levulinoyl-beta-D-galactopyranosyl-(1-->4)-2,3,6-tri-O-benzyl-beta-D-galactopyranosyl-(1-->4)-2,3,6-tri-O-benzyl-beta-D-glucopyranoside and 2-(trimethylsilyl)ethyl 2-acetamido-2-deoxy-6-O-levulinoyl-beta-D-galactopyranosyl-(1-->4)-2,6-di-O-benzyl-3-O-levulinoyl-beta-D-galactopyranosyl-(1-->4)-2,3,6-tri-O-benzyl-beta-D-glucopyranoside were each glycosylated with alpha-NeuAc-(2-->3)-galactose

Articles

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service